Skip to Content
Merck
All Photos(1)

Key Documents

22197

Supelco

Cellulose

DS-0, powder, suitable for thin layer chromatography (TLC)

Synonym(s):

Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41115711
NACRES:
NC.07

Quality Level

form

powder

technique(s)

thin layer chromatography (TLC): suitable

ign. residue

≤0.1%

loss

≤5% loss on drying

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose, a homopolymer consisting of glucose units joined by β-1,4 bonds. These are strongly attached through inter and intramolecular hydrogen bonds and vander Waals forces resulting in microfibrils which together form fibers. They are arranged in parallel reducing ends of adjacent glucan chains. Cellulose is widely used as sorbent in TLC.

Application

Cellulose was used to measure the cyrstallinity index using X-ray diffraction (XRD) patterns and solid-state 13C nuclear magnetic resonance (NMR) spectra.
High purity cellulose powders for partition chromatography.
microcrystalline cellulose for all types of separation chromatography and thin layer electrophoresis

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jadranka Odović et al.
Iranian journal of pharmaceutical research : IJPR, 11(3), 763-770 (2012-07-01)
In this assay, the evaluation of lipophilicity of four ACE-inhibitors and hydrochlorothiazide (HCTZ) with RP-TLC on cellulose layers was described using three binary solvent systems. The selected ACE inhibitors had sufficiently different structures which can indicate the method suitability for
Sunkyu Park et al.
Biotechnology for biofuels, 3, 10-10 (2010-05-26)
Although measurements of crystallinity index (CI) have a long history, it has been found that CI varies significantly depending on the choice of measurement method. In this study, four different techniques incorporating X-ray diffraction and solid-state 13C nuclear magnetic resonance
S B Leschine
Annual review of microbiology, 49, 399-426 (1995-01-01)
In anaerobic environments rich in decaying plant material, the decomposition of cellulose is brought about by complex communities of interacting microorganisms. Because the substrate, cellulose, is insoluble, bacterial and fungal degradation occurs exocellularly, either in association with the outer cell
Ginger Brininstool et al.
BMC plant biology, 8, 58-58 (2008-05-20)
The Arabidopsis thaliana CONSTITUTIVE EXPRESSOR OF PATHOGENESIS-RELATED GENES5 (CPR5) gene has been previously implicated in disease resistance, cell proliferation, cell death, and sugar sensing, and encodes a putative membrane protein of unknown biochemical function. Trichome development is also affected in
Jung Min Lee et al.
JAMA otolaryngology-- head & neck surgery, 139(6), 629-635 (2013-06-22)
Bioresorbable fixation systems have been popular for the treatment of facial fractures. However, their mechanical properties are uncertain and complications have been reported. To overcome these problems, we developed a bioresorbable fixation plate using a composite of silk fibroin and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service