Skip to Content
Merck
All Photos(1)

Key Documents

22182

Supelco

Cellulose Chromatography Resin

acid washed, from spruce, 100-700 mesh

Synonym(s):

Cellulose powder, Cotton linters

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
23151817
NACRES:
SB.52

Product Name

Cellulose, acid washed, from spruce, for column chromatography

biological source

spruce

Quality Level

grade

for column chromatography

form

powder

quality

acid washed

technique(s)

LPLC: suitable

fiber L

0.02-0.15 mm

impurities

≤0.05% ether extract

ign. residue

~0.01%

matrix

Cellulose

matrix active group

polymer

particle size

20-150 μm

cation traces

Cu: ≤5 mg/kg
Fe: ≤10 mg/kg

suitability

suitable for column chromatography

separation technique

size exclusion (SEC)

SMILES string

O1[C@H](C(C(C(C1CO)O)O)O)O[C@@H]2C(OC(C(C2O)O)O)CO

InChI

1S/C12H22O11/c13-1-3-5(15)6(16)9(19)12(22-3)23-10-4(2-14)21-11(20)8(18)7(10)17/h3-20H,1-2H2/t3?,4?,5?,6?,7?,8?,9?,10-,11?,12+/m1/s1

InChI key

GUBGYTABKSRVRQ-WFVLMXAXSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cellulose, a homopolymer consisting of glucose units joined by beta-1,4 bonds. These are strongly attached through inter and intramolecular hydrogen bonds and van der Waals forces resulting in microfibrils which together form fibers. They are arranged in parallel reducing ends of adjacent glucan chains. Cellulose is widely used as sorbent in TLC.

Application

Cellulose was used to measure the cyrstallinity index using X-ray diffraction (XRD) patterns and solid-state 13C nuclear magnetic resonance spectra.
High purity cellulose powders for partition chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marco Mattonai et al.
Bioresource technology, 270, 270-277 (2018-09-18)
A combined study of crystallinity index (CI), degree of polymerisation (DP) and thermal stability of cellulose was carried out for monitoring the effect of ball-milling. DP and CI are two fundamental quantities that describe the physico-chemical behaviour of cellulose. Milling
Jadranka Odović et al.
Iranian journal of pharmaceutical research : IJPR, 11(3), 763-770 (2012-07-01)
In this assay, the evaluation of lipophilicity of four ACE-inhibitors and hydrochlorothiazide (HCTZ) with RP-TLC on cellulose layers was described using three binary solvent systems. The selected ACE inhibitors had sufficiently different structures which can indicate the method suitability for
S B Leschine
Annual review of microbiology, 49, 399-426 (1995-01-01)
In anaerobic environments rich in decaying plant material, the decomposition of cellulose is brought about by complex communities of interacting microorganisms. Because the substrate, cellulose, is insoluble, bacterial and fungal degradation occurs exocellularly, either in association with the outer cell
Sunkyu Park et al.
Biotechnology for biofuels, 3, 10-10 (2010-05-26)
Although measurements of crystallinity index (CI) have a long history, it has been found that CI varies significantly depending on the choice of measurement method. In this study, four different techniques incorporating X-ray diffraction and solid-state 13C nuclear magnetic resonance
Ginger Brininstool et al.
BMC plant biology, 8, 58-58 (2008-05-20)
The Arabidopsis thaliana CONSTITUTIVE EXPRESSOR OF PATHOGENESIS-RELATED GENES5 (CPR5) gene has been previously implicated in disease resistance, cell proliferation, cell death, and sugar sensing, and encodes a putative membrane protein of unknown biochemical function. Trichome development is also affected in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service