21572
Hamilton® syringe, 700 series, special needle
701SN, volume 10 μL, needle size 26s ga (bevel tip), needle L 76 mm (3 in.)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
Recommended Products
material
stainless steel needle (point style 2)
needle size
26s ga (bevel tip)
feature
needle type SN (special needle)
packaging
pkg of 1 ea
manufacturer/tradename
Hamilton 80384
needle L
76 mm (3 in.)
volume
10 μL
compatibility
for use with Hamilton 701SN (special needle)
Looking for similar products? Visit Product Comparison Guide
General description
Hamilton syringe is available in gas-tight type that minimizes the gas exchange in GC analysis. It gives accurate volume delivery. The plunger is mostly unsurpassed.
Application
It has been used in the extraction procedure of urine samples for analysing cocaine and cocaine metabolites using hollow fibre membrane solvent microextraction (HFMSME).
Legal Information
Hamilton is a registered trademark of Hamilton Co.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Preliminary studies of a fast screening method for cocaine and cocaine metabolites in urine using hollow fibre membrane solvent microextraction (HFMSME).
de Jager, Lowri S., and Anthony RJ Andrews.
Analyst, 126.8, 1298-1303 (2001)
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Wolf, Don P.
In Vitro Fertilization and Embryo Transfer: A Manual of Basic Techniques, 06-Dec, 192-192 (2012)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service