Skip to Content
Merck
All Photos(1)

Documents

20370

Sigma-Aldrich

tert-Butyl nitrite

technical, ≥90% (GC)

Synonym(s):

1,1-Dimethylethyl nitrite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3CONO
CAS Number:
Molecular Weight:
103.12
Beilstein:
1209339
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Quality Level

Assay

≥90% (GC)

form

liquid

impurities

≤8% tert-butyl alcohol

refractive index

n20/D 1.368 (lit.)
n20/D 1.368

bp

61-63 °C (lit.)

solubility

alcohol: very soluble
chloroform: very soluble
diethyl ether: very soluble
glycerol: insoluble
water: slightly soluble

density

0.864 g/mL at 20 °C
0.867 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CC(C)(C)ON=O

InChI

1S/C4H9NO2/c1-4(2,3)7-5-6/h1-3H3

InChI key

IOGXOCVLYRDXLW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

tert-Butyl nitrite is an organic building block widely used in various organic transformations such as oxidation, diazotization, nitrosation, nitration, oximation, N-synthon, and miscellaneous reactions.

Application

tert-Butyl nitrite can be used as a reactant to synthesize aryl diazonium salts via diazotation.

Other Notes

Reagent for nitrosations, kinetics; Clean source of Me3C radicals by near-UV photolysis

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

14.0 °F - closed cup

Flash Point(C)

-10 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantum defects as versatile anchors for carbon nanotube functionalization
Mann F, et al.
Nature Protocols, 17, 727-747 (2022)
M.J. Crookes et al.
J. Chem. Soc. Perkin Trans. II, 1339-1339 (1988)
tert-Butyl Nitrite (TBN), a Multitasking Reagent in Organic Synthesis
Dahiya A, et al.
Chemistry - An Asian Journal, 14, 4454-4492 (2019)
Brenden Kilpatrick et al.
Chemical communications (Cambridge, England), 49(5), 514-516 (2012-12-01)
A methodology for the efficient conversion of aromatic sulfonamides into their mono-nitro derivatives using tert-butyl nitrite is reported. The reaction exhibits a high degree of chemoselectivity for sulfonamide functionalized aryl systems, even in the presence of other sensitive or potentially
L. Batt et al.
International Journal of Chemical Kinetics, 21, 535-535 (1989)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service