Skip to Content
Merck
All Photos(3)

Documents

134236

Sigma-Aldrich

Morpholine

ReagentPlus®, ≥99%

Synonym(s):

Tetrahydro-1,4-oxazine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H9NO
CAS Number:
Molecular Weight:
87.12
Beilstein:
102549
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39162409
PubChem Substance ID:
NACRES:
NA.21

vapor density

3 (vs air)

Quality Level

vapor pressure

31 mmHg ( 38 °C)
7 mmHg ( 20 °C)

product line

ReagentPlus®

Assay

≥99%

form

liquid

autoignition temp.

590 °F

expl. lim.

10.8 %

refractive index

n20/D 1.454 (lit.)

bp

129 °C (lit.)

mp

−7-−5 °C (lit.)

density

0.996 g/mL at 25 °C (lit.)

SMILES string

C1COCCN1

InChI

1S/C4H9NO/c1-3-6-4-2-5-1/h5H,1-4H2

InChI key

YNAVUWVOSKDBBP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Morpholine can be obtained, via degradation of N-methylmorpholine N-oxide (NMMO) during fiber production.

Morpholine is a cyclic amine that serves as a solvent, corrosion inhibitor, and chemical intermediate.

Application

Morpholine can be used as a buffer component in the separation of peptides and pathogenic lipopolysaccharides during on-line chromatographic preconcentration coupled to capillary zone electrophoresis–electrospray mass spectrometry (cPC–CZE–ES-MS) analysis from colony isolates.

Legal Information

ReagentPlus is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Repr. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

87.8 °F - closed cup

Flash Point(C)

31 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Flow injection analysis with amperometric detection for morpholine determination in corrosion inhibitors
Oliveira SMD, et al.
Journal of the Brazilian Chemical Society, 25, 1399-1406 (2014)
"Determination of morpholine and N-methylmorpholine formed as degradation products in cellulose fiber production by capillary isotachophoresis"
Widhalm A and Kenndler E
Analytical Chemistry, 63(06), 645-648 (1991)
"Development of an on-line preconcentration method for the analysis of pathogenic lipopolysaccharides using capillary electrophoresis?electrospray mass spectrometry: Application to small colony isolates1NRCC No.: 39585.1"
Li.J, et al.
Journal of Chromatography A, 817(01), 325-336 (1998)
Nickel-catalyzed amination of aryl sulfamates.
Stephen D Ramgren et al.
Angewandte Chemie (International ed. in English), 50(9), 2171-2173 (2011-02-24)
M H Jayan S Karunarathna et al.
Photochemical & photobiological sciences : Official journal of the European Photochemistry Association and the European Society for Photobiology, 20(2), 255-263 (2021-03-16)
The photochemistry of Fe(III) coordinated to natural uronate-containing polysaccharides has been investigated quantitatively in aqueous solution. It is demonstrated that the photoreduction of the coordinated Fe(III) to Fe(II) and oxidative decarboxylation occurs in a variety of uronate-containing polysaccharides. The photochemistry

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service