Skip to Content
Merck
All Photos(4)

Key Documents

12806

Sigma-Aldrich

O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate

≥97.0% (N), for peptide synthesis

Synonym(s):

N,N,N′,N′-Tetramethyl-O-(benzotriazol-1-yl)uronium tetrafluoroborate, TBTU

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C11H16N5O · BF4
CAS Number:
Molecular Weight:
321.08
Beilstein:
7066325
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.21

Product Name

O-(Benzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium tetrafluoroborate, ≥97.0% (N)

Quality Level

Assay

≥97.0% (N)

form

solid

mp

205 °C (dec.) (lit.)

solubility

acetonitrile: 0.1 g/mL, clear, colorless

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

F[B-](F)(F)F.CN(C)\C(On1nnc2ccccc12)=[N+](/C)C

InChI

1S/C11H16N5O.BF4/c1-14(2)11(15(3)4)17-16-10-8-6-5-7-9(10)12-13-16;2-1(3,4)5/h5-8H,1-4H3;/q+1;-1

InChI key

JKEKMBGUVUKMQB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Coupling reagent which is ideally suited for solid-phase peptide synthesis.

Other Notes

Recent studies show that crystal as well as solution structure of this coupling agent is a guanidinium N-oxide and not an uronium compound; Coupling reagent which is ideally suited for solid-phase peptide synthesis.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Sol. 1 - Skin Sens. 1A

Storage Class Code

4.1A - Other explosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rajni Bala et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 196, 268-273 (2018-02-20)
In the present study, we report a highly sensitive, rapid and low cost colorimetric monitoring of malathion (an organophosphate insecticide) employing a basic hexapeptide, malathion specific aptamer (oligonucleotide) and silver nanoparticles (AgNPs) as a nanoprobe. AgNPs are made to interact
S. Zimmer et al. et al.
Proc. 22nd Eur. Pept. Symp.: Peptides 1992 null
Parvin Zakeri-Milani et al.
EXCLI journal, 16, 650-662 (2017-07-12)
Gemcitabine (Gem) is used as a single agent or in combination with other anticancer agents to treat many types of solid tumors. However, it has many limitations such as a short plasma half-life, dose-limiting toxicities and drug resistance. Cell-penetrating peptides
O-(BENZOTRIAZOL-1-YL)-N, N, N', N'-TETRAMETHYLURONIUM TETRAFLUOROBORATE (TBTU): A NEW REAGENT FOR THE CLEAVAGE OF TETRAHYDROPYRANYL, SILYL AND 4, 4'-D IMETHOXYTRITYL ETHERS.
Ramasamy KS and Averett D.
Synlett, 6, 709-712 (1999)
I. Abdelmoty
Lett. Pept. Sci., 1, 57-57 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service