Skip to Content
Merck
All Photos(1)

Key Documents

12669

Sigma-Aldrich

Potassium thiosulfate

≥95%

Synonym(s):

Dipotassium thiosulfate, Thiosulfuric acid (H2S2O3), potassium salt (1:2), Thiosulfuric acid, dipotassium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2S2O3
CAS Number:
Molecular Weight:
190.32
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22
grade:
for analytical purposes
form:
solid

grade

for analytical purposes

Quality Level

Assay

≥95%

form

solid

SMILES string

[K+].[K+].[O-]S([O-])(=O)=S

InChI

1S/2K.H2O3S2/c;;1-5(2,3)4/h;;(H2,1,2,3,4)/q2*+1;/p-2

InChI key

FGRVOLIFQGXPCT-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Potassium thiosulfate can be used:
  • For the preparation of aqueous biphasic systems (ABS) for the extraction of organic compounds.
  • For the synthesis of polyacrylonitrile nanoparticles by dispersion/emulsion polymerization.
  • As a reagent for the pretreatment of graphite powder to synthesize graphene oxide from it by Hummers′ method.
  • As an initiator to synthesize organosilicon-based adsorbent to remove iron from crude phosphoric acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ying Chen et al.
Ecotoxicology and environmental safety, 189, 109929-109929 (2019-11-14)
Sewage treatment plants effectively remove silver (Ag) from sewage. Sewage sludge can therefore be important Ag sinks, polluting the environment with this element. In this work, we report a nation-wide survey on the Ag content of sewage sludge in China
C T Perciani et al.
Journal of clinical laboratory analysis, 21(3), 201-206 (2007-05-18)
The methods currently used to determine the immunoglobulin avidity index (AI) require the choice of a reference point in the ELISA titration curve. Since both curves, with and without denaturating reagents, seldom run in parallel, the AI value becomes highly
Preparation of Cu-graphene coating via electroless plating for high mechanical property and corrosive resistance.
Li S, et al.
Journal of alloys and compounds, 777(2), 877-885 (2019)
Ruijun Qin et al.
Chemosphere, 72(4), 558-563 (2008-04-29)
Plastic tarps are commonly used in raised bed strawberry production to minimize emissions of preplant soil fumigants and are left in place throughout the growing season as part of the standard cultural practices. Soil amendments with chemicals such as thiosulfate
Chao Hou et al.
Physical chemistry chemical physics : PCCP, 19(35), 24126-24134 (2017-08-25)
A multilayered architecture including a thin Au film supporting an X-shaped nanohole array and a thick continuous Au film separated by a Cytop dielectric layer is reported in this work. Long-range surface plasmon resonance (LR-SPR) was generated at the top

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service