Skip to Content
Merck
All Photos(1)

Key Documents

12363

Sigma-Aldrich

Iron(II) sulfide

sticks (thin)

Synonym(s):

Ferrous sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
FeS
CAS Number:
Molecular Weight:
87.91
EC Number:
MDL number:
UNSPSC Code:
12352302
eCl@ss:
38190405
PubChem Substance ID:
NACRES:
NA.21
form:
sticks (thin)

form

sticks (thin)

Quality Level

concentration

~25% S

density

4.84 g/mL at 25 °C (lit.)

SMILES string

S=[Fe]

InChI

1S/Fe.S

InChI key

MBMLMWLHJBBADN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Effects of solution composition and pH on the reductive dechlorination of hexachloroethane by iron sulfide.
Butler EC and Hayes KF.
Environmental Science & Technology, 32(9), 1276-1284 (1998)
Oxidation of pyrite and iron sulfide by manganese dioxide in marine sediments.
Schippers A and J?rgensen BB.
Geochimica et Cosmochimica Acta, 65(6), 915-922 (2001)
Elisa Guazzelli et al.
Polymers, 12(6) (2020-06-04)
Diblock copolymers composed of a polystyrene first block and a PEG-fluoroalkyl chain-modified polystyrene second block were synthesized by controlled atom transfer radical polymerization (ATRP), starting from the same polystyrene macroinitiator. The wettability of the polymer film surfaces was investigated by
N Finck et al.
Environmental science & technology, 46(18), 10004-10011 (2012-08-21)
The isotope (79)Se may be of great concern with regard to the safe disposal of nuclear wastes in deep geological repositories due to its long half-life and potential mobility in the geosphere. The Se mobility is controlled by the oxidation
Amit Kumar Dutta et al.
ACS applied materials & interfaces, 4(4), 1919-1927 (2012-03-28)
Nanocrystalline FeS and FeSe compounds were prepared by solvothermal decomposition of a precursor complex [Fe(3)(μ(3)-O)(μ(2)-O(2)CCH(2)Cl)(6)(H(2)O)(3)]NO(3)·H(2)O in the presence of thiourea and sodium selenite, respectively. The as-obtained products were characterized by X-ray diffraction analysis (XRD), field emission scanning electron microscopy (FESEM)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service