Skip to Content
Merck
All Photos(1)

Key Documents

08299

Supelco

Cyclopentanone

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H8(=O)
CAS Number:
Molecular Weight:
84.12
Beilstein:
605573
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥99.5% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.437 (lit.)
n20/D 1.437

bp

130-131 °C (lit.)

mp

−51 °C (lit.)

density

0.951 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

neat

SMILES string

O=C1CCCC1

InChI

1S/C5H8O/c6-5-3-1-2-4-5/h1-4H2

InChI key

BGTOWKSIORTVQH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclopentanone is one of the industrially important compounds employed in the synthesis of fungicides, pharmaceuticals, rubber chemicals and perfume products.

Application

Cyclopentanone may be used as a reference standard for the determination of cyclopentanone in:
  • Pyrolysis oil samples by gas chromatography with flame ionization detector (GC-FID) as well as two-dimensional GC with time-of-flight mass spectrometry (GC×GC-TOFMS).
  • Brazilian sugarcane spirits and rum by high-performance liquid chromatography (HPLC) with ultraviolet-visible (UV-Vis) diode array detector (DAD).
  • Crude and hydrotreated bio-oils obtained from various biomass resources by GC×GC-FID as well as GC×GC-TOF MS.
  • Indoor air samples of semiconductor foundries by multisorbent adsorption/thermal desorption combined with GC-MS.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

86.0 °F - closed cup

Flash Point(C)

30 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qualitative and quantitative analysis of pyrolysis oil by gas chromatography with flame ionization detection and comprehensive two-dimensional gas chromatography with time-of-flight mass spectrometry
Sfetsas T, et al.
Journal of Chromatography A, 1218(21), 3317-3325 (2011)
Effect of catalyst and solvent on the furan ring rearrangement to cyclopentanone
Hronec M, et al.
Applied Catalysis A: General, 437, 104-111 (2012)
Quantitative analysis of crude and stabilized bio-oils by comprehensive two-dimensional gas-chromatography
Djokic MR, et al.
Journal of Chromatography A, 1257(5), 131-140 (2012)
Catalytic vapor-phase cyclization of 1, 6-hexanediol into cyclopentanone
Akashi T, et al.
Catalysis Communications, 4(8), 411-416 (2003)
Measurement of toxic volatile organic compounds in indoor air of semiconductor foundries using multisorbent adsorption/thermal desorption coupled with gas chromatography-mass spectrometry
Wu CH, et al.
Journal of Chromatography A, 996(1-2), 225-231 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service