Skip to Content
Merck
All Photos(1)

Key Documents

05670

Sigma-Aldrich

Allantoin

≥98.0% (N)

Synonym(s):

NSC 7606, 5-Ureidohydantoin, Glyoxylic(acid) diureide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N4O3
CAS Number:
Molecular Weight:
158.12
Beilstein:
102364
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (N)

form

powder

ign. residue

≤0.2%

mp

230 °C (dec.) (lit.)

functional group

amine

SMILES string

NC(=O)NC1NC(=O)NC1=O

InChI

1S/C4H6N4O3/c5-3(10)6-1-2(9)8-4(11)7-1/h1H,(H3,5,6,10)(H2,7,8,9,11)

InChI key

POJWUDADGALRAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for:
Synthesis of Mannich bases for use as antibacterial agents
Synthesis of sulofenur analogs for use as antitumor agents

Biochem/physiol Actions

Purine metabolite via the uric acid pathway. Uric acid also reacts with free radicals to produce allantoin, thus allantoin may be a useful biomarker for oxidative stress.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sh. Joshi
Oxidation Communications, 32, 714-723 (2009)
Khairia M. Youssef, et al
Medicinal Chemistry Research, 11, 481-503 (2003)
Peng Wang et al.
Journal of agricultural and food chemistry, 60(11), 2793-2798 (2012-03-01)
Despite increasing knowledge of allantoin as a phytochemical involved in rice, relatively little is known about its distribution and function in rice grains. In this study, allantoin was quantified in 15 Chinese rice grains, and its contents varied with grain
Camila G Moreira et al.
Journal of ethnopharmacology, 141(3), 1005-1011 (2012-04-17)
Pyrostegia venusta is a native Brazilian plant which has a variety of uses in traditional folk medicine including the treatment of vitiligo. However, its effectiveness on melanogenesis is not yet elucidated. This study aimed to investigate the melanogenic activity of
Dora Il'yasova et al.
Clinica chimica acta; international journal of clinical chemistry, 413(19-20), 1446-1453 (2012-06-12)
Oxidative damage produced by reactive oxygen species (ROS) has been implicated in the etiology and pathology of many health conditions, including a large number of chronic diseases. Urinary biomarkers of oxidative status present a great opportunity to study redox balance

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service