05419
Germanium Standard for ICP
TraceCERT®, 1 g/L Ge in nitric acid (nominal concentration)
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
certified reference material
TraceCERT®
Quality Level
product line
TraceCERT®
concentration
1 g/L Ge in nitric acid (nominal concentration)
technique(s)
ICP: suitable
application(s)
industrial qc
pharmaceutical
format
single component solution
General description
This certified reference material (CRM) is produced in accordance with ISO 17034 and characterized in accordance with ISO/IEC 17025. This CRM is traceable to SI unit kg and measured against primary material from a National Metrology Institute (NMI), e.g. NIST.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at https://www.sigma-aldrich.comto view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Please visit ISO certificates and Site Quality Self-Assessments to access the current certificates of accreditation.
Download your certificate at https://www.sigma-aldrich.comto view certified values, including uncertainty, date of expiry, and detailed information about trace impurities.
Application
Germanium ICP standard can be used as a calibration standard in the determination of isotopic compositions of germaniumin sulfide minerals by inductively coupled plasma mass spectrometry (ICP-MS).
Analysis Note
1g/L Ge in 2% nitric acid (contains HF traces), prepared with high purity Ge metal, HNO3, HF and water
Other Notes
For a complete product listing of our TraceCERT® range of CRMs for ICP and AAS, technical information and example certificates please visit our ICP& AAS standards website
Legal Information
TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Veterinary research, 52(1), 4-4 (2021-01-09)
The transport of circovirus capsid protein into nucleus is essential for viral replication in infected cell. However, the role of nucleolar shuttle proteins during porcine circovirus 3 capsid protein (PCV3 Cap) import is still not understood. Here, we report a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service