Skip to Content
Merck
All Photos(1)

Key Documents

04936

Sigma-Aldrich

O-(6-Chlorobenzotriazol-1-yl)-N,N,N′,N′-tetramethyluronium hexafluorophosphate

≥98.0% (HPLC)

Synonym(s):

1-[Bis(dimethylamino)methylen]-5-chlorobenzotriazolium 3-oxide hexafluorophosphate, N,N,N′,N′-Tetramethyl-O-(6-chloro-1H-benzotriazol-1-yl)uronium hexafluorophosphate, HCTU

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H15ClF6N5OP
CAS Number:
Molecular Weight:
413.69
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98.0% (HPLC)

form

powder

reaction suitability

reaction type: Coupling Reactions

impurities

≤0.5% water

mp

185-190 °C

application(s)

peptide synthesis

functional group

amine
chloro

SMILES string

ClC1=CC=C(N([C+](N(C)C)N(C)C)N=[N+]2[O-])C2=C1.F[P-](F)(F)(F)(F)F

InChI

1S/C11H15ClN5O.F6P/c1-15(2)11(16(3)4)18-17-10-7-8(12)5-6-9(10)13-14-17;1-7(2,3,4,5)6/h5-7H,1-4H3;/q+1;-1

InChI key

ZHHGTMQHUWDEJF-UHFFFAOYSA-N

Application

Reagent for:
Synthesis of near-infrared pH activatable fluorescent probes
Synthesis of human β-amyloid by Fmoc chemistry
Stereoselective Horner-Wadsworth-Emmons olefination
Covalent ligation of fluorescent peptides to quantum dots
Alkylation of human telomere sequence

Other Notes

Tandem Oligonucleotide Synthesis on Solid-Phase Supports for the Production of Multiple Oligonucleotides

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1A

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Juan B Blanco-Canosa et al.
Journal of the American Chemical Society, 132(29), 10027-10033 (2010-07-06)
Water solubilized nanoparticles such as CdSe-ZnS core-shell nanocrystals (quantum dots, QDs) have great potential in bioimaging and sensing applications due to their excellent photophysical properties. However, the efficient modification of QDs with complex biomolecules represents a significant challenge. Here, we
Wouter A van der Linden et al.
Organic & biomolecular chemistry, 8(8), 1885-1893 (2010-05-08)
The synthesis and biological evaluation of ten Michael acceptors containing potential proteasome inhibitors are described. Cellular targets of azide containing inhibitors and were assessed in HEK293T and RAW264.7 cells by a two step labeling strategy, followed by biotin-pulldown, affinity purification
Fast conventional synthesis of human beta-amyloid (1-42) on the Symphony and Prelude.
German Fuentes et al.
Advances in experimental medicine and biology, 611, 173-174 (2009-04-30)
Amit Kumar Tripathi et al.
Scientific reports, 7(1), 3384-3384 (2017-06-15)
Marine fish antimicrobial peptide, chrysophsin-1 possesses versatile biological activities but its non-selective nature restricts its therapeutic possibilities. Often small alterations in structural motifs result in significant changes in the properties of concerned proteins/peptides. We have identified GXXXXG motif in chrysophsin-1.
Hyeran Lee et al.
Bioconjugate chemistry, 22(4), 777-784 (2011-03-11)
Highly tumor selective near-infrared (NIR) pH-activatable probe was developed by conjugating pH-sensitive cyanine dye to a cyclic arginine-glycine-aspartic acid (cRGD) peptide targeting α(v)β(3) integrin (ABIR), a protein that is highly overexpressed in endothelial cells during tumor angiogenesis. The NIR pH-sensitive

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service