Skip to Content
Merck
All Photos(1)

Key Documents

04899

Supelco

α-Terpineol

analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H18O
CAS Number:
Molecular Weight:
154.25
Beilstein:
2325137
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24
Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥95.0% (GC)

shelf life

limited shelf life, expiry date on the label

impurities

≤1.0% water

refractive index

n20/D 1.482 (lit.)

bp

217-218 °C (lit.)

mp

31-35 °C (lit.)

density

0.93 g/mL at 25 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

CC1=CC[C@H](CC1)C(C)(C)O

InChI

1S/C10H18O/c1-8-4-6-9(7-5-8)10(2,3)11/h4,9,11H,5-7H2,1-3H3

InChI key

WUOACPNHFRMFPN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Terpineol is a monoterpene alcohol. It is one of the components responsible for the antifungal activity of Melaleuca alternifolia (tea tree) essential oil.[1] The reaction rate constant of α-terpineol with OH radical and ozone was found to be (1.9±0.5)×10-10cm3 molecule-1s-1 and (3.0±0.2)×10-16cm3 molecule-1s-1, respectively.[2]

Application

α-Terpineol has been used as a reference standard for the identification of α-terpineol in tequila by solid phase microextraction (SPME) and capillary gas chromatography−mass spectrometry (GC-MS),[3] and in virgin olive oil samples by headspace SPME followed by GC-MS equipped with flame ionization detector (FID).[4]
It may be used as a reference standard for the analysis of α-terpineol as trace volatile compound in wine by solid-phase extraction (SPE) followed by GC-MS[5] and in essential oils and related compounds from aromatic plants by SPME and GC-MS.[6]
α-Terpineol may be used in the synthesis of α-terpinyl esters of acetic acid and acetic anhydride via lipase-mediated esterification.[7]

Other Notes

This compound is commonly found in plants of the genus: cinnamomum mentha pimpinella salvia thymus valeriana glycyrrhiza
Depending on the ambient or transport temperature the aggregation state can vary.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

195.8 °F - closed cup

Flash Point(C)

91 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of terpenes in tequila by solid phase microextraction-gas chromatography-mass spectrometry.
Pe?a-Alvarez, et al.
Journal of Chromatography A, 1134(1-2), 291-297 (2006)
Jana Richter et al.
Analytical and bioanalytical chemistry, 387(6), 2207-2217 (2007-01-16)
Different extraction methods for the subsequent gas chromatographic determination of the composition of essential oils and related compounds from marjoram (Origanum majorana L.), caraway (Carum carvi L.), sage (Salvia officinalis L.), and thyme (Thymus vulgaris L.) have been compared. The
Determination of minor and trace volatile compounds in wine by solid-phase extraction and gas chromatography with mass spectrometric detection.
Lopez, et al.
Journal of Chromatography A, 966(1-2), 167-177 (2002)
Lipase catalyzed esterification of a-terpineol with various organic acids: application of the Plackett?Burman design.
Rao P and Divakar S.
Process. Biochem., 36(11), 1125-1128 (2001)
Antifungal activity of the components of Melaleuca alternifolia (tea tree) oil.
Hammer KA , et al.
Journal of Applied Microbiology, 95(4), 853-860 (2003)

Protocols

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

HPTLC fingerprinting enables rapid identification of passion flower with related reference materials.

See All

Questions

  1. What is the density of Alpha Terpineol?

    1 answer
    1. 0.93 g/mL at 25 °C (lit.)

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service