Skip to Content
Merck
All Photos(1)

Key Documents

04700

Sigma-Aldrich

4-Ethylphenol

≥97.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5C6H4OH
CAS Number:
Molecular Weight:
122.16
Beilstein:
1363317
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39023356
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor density

4.2 (vs air)

Quality Level

vapor pressure

0.13 mmHg ( 20 °C)

Assay

≥97.0% (GC)

bp

218-219 °C (lit.)

mp

40-42 °C (lit.)
41-44 °C

SMILES string

CCc1ccc(O)cc1

InChI

1S/C8H10O/c1-2-7-3-5-8(9)6-4-7/h3-6,9H,2H2,1H3

InChI key

HXDOZKJGKXYMEW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Eduardo Chicote et al.
Journal of industrial microbiology & biotechnology, 32(4), 155-162 (2005-03-22)
The aim of the present research was to isolate and identify bacteria from spent nuclear fuel pools of a Spanish nuclear power plant. Water samples were collected and inoculated onto different culture media to isolate the highest number of species.
C van Zyl et al.
Applied and environmental microbiology, 59(5), 1487-1494 (1993-05-01)
The influence of D-ribose as a cosubstrate on the uptake and metabolism of the non-growth substrate D-xylose by Saccharomyces cerevisiae ATCC 26602 was investigated. Xylose was taken up by means of low- and high-affinity glucose transport systems. In cells exposed
Etsuro Matsubara et al.
Journal of neurochemistry, 85(5), 1101-1108 (2003-05-20)
Increased levels of a 40-42 amino-acid peptide called the amyloid beta protein (A beta) and evidence of oxidative damage are early neuropathological markers of Alzheimer's disease (AD). Previous investigations have demonstrated that melatonin is decreased during the aging process and
S Shivaji et al.
International journal of systematic and evolutionary microbiology, 59(Pt 12), 2977-2986 (2009-08-01)
Three novel bacterial strains, PVAS-1(T), B3W22(T) and B8W22(T), were isolated from cryotubes used to collect air samples at altitudes of between 27 and 41 km. Based on phenotypic characteristics, chemotaxonomic features, DNA-DNA hybridization with the nearest phylogenetic neighbours and phylogenetic
Pamela Y Quintas et al.
Archives of environmental contamination and toxicology, 81(2), 307-323 (2021-07-17)
This study evaluates for the first time the distribution and accumulation of butyltin compounds (BTs) in different compartments such as seawater, sediments, suspended particulate matter (SPM), and mussels (Brachidontes rodriguezii) in the Bahía Blanca estuary. The samples were collected from

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service