Skip to Content
Merck
All Photos(1)

Key Documents

04078

Sigma-Aldrich

Hydrogen peroxide–Urea adduct

powder, 15-17% active oxygen basis

Synonym(s):

Carbamide Per hydrate, Carbamide peroxide, Percarbamide, Urea hydrogen peroxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CO(NH2)2 · H2O2
CAS Number:
Molecular Weight:
94.07
Beilstein:
3680414
EC Number:
MDL number:
UNSPSC Code:
12352111
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

23.3 mmHg ( 30 °C)

Quality Level

form

powder

composition

active oxygen, 15-17%

reaction suitability

reagent type: oxidant

mp

85-90 °C (dec.)
90-93 °C (lit.)

functional group

amine

SMILES string

OO.NC(N)=O

InChI

1S/CH4N2O.H2O2/c2-1(3)4;1-2/h(H4,2,3,4);1-2H

InChI key

AQLJVWUFPCUVLO-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Hydrogen peroxide–Urea adduct is a compound composed of hydrogen peroxide (H2O2) and urea (CO(NH2)2) in equal amount. UHP, also known as carbamide peroxide, is manufactured industrially as a solid source of hydrogen peroxide for bleaching, disinfection, and oxidation reactions.

Application

Hydrogen peroxide–Urea adduct is used as an oxidizing agent.
Reactant which undergoes solid-state decomposition as a source of peroxide

Reactant used as an oxidant

Features and Benefits

Hydrogen peroxide–Urea (HPU) adduct is

  • Inexpensive
  • Commercially available
  • Stable solid.

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 3 - Skin Irrit. 2

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Highly efficient epoxidation of propylene to propylene oxide over TS-1 using urea+ hydrogen peroxide as oxidizing agent
Cheng W, et al.
Chinese Journal of Catalysis, 255(2) , 343-346 (2008)
Small-Scale Detonation of Industrial Urea-Hydrogen Peroxide (UHP )
Halleux F, et al.
Propellants, Explosives, Pyrotechnics, 47(2), e202100250-e202100250 (2022)
T. Michel, et al.,
J. Mol. Catal. A: Chem., 340, 9-14 (2011)
N. O. Mahmoodi, et al.,
Synthetic Communications, 40, 3181-3185 (2010)
Solvent, metal and halogen-free synthesis of sulfoxides by using a recoverable heterogeneous urea-hydrogen peroxide silica-based oxidative catalytic system
Azizi M, et al.
Catalysis Communications, 100, 62-65 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service