01986
(±)-β-Hexabromocyclododecane
analytical standard
Synonym(s):
(1RS,2SR,5RS,6RS,9RS,10SR)-1,2,5,6,9,10-Hexabromocyclododecane, (±)-β-HBCD
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C12H18Br6
CAS Number:
Molecular Weight:
641.70
UNSPSC Code:
41116107
NACRES:
NA.24
Recommended Products
grade
analytical standard
Quality Level
Assay
≥97.0% (HPLC)
shelf life
limited shelf life, expiry date on the label
impurities
≤1.0% water
mp
152-158 °C
format
neat
Related Categories
General description
β-Hexabromocyclododecane is the minor diastereomer in the production of the brominated flame retardant (BFR), hexabromocyclododecane (HBCD). It contributes to 16% of the total mixture. HBCD is widely used in a variety of commercial and consumer products.
Application
β-Hexabromocyclododecane may be used as an analytical reference standard for the determination of the analyte in textiles, air, and soil, and various other samples by chromatography techniques.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1 - Lact. - Repr. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Determination of flame-retardant hexabromocyclododecane diastereomers in textiles
Kajiwara N, et al.
Chemosphere, 74(11), 1485-1489 (2009)
Dietary intake of hexabromocyclododecane diastereoisomers ($\alpha$-, $\beta$-, and $\gamma$-HBCD) in the Belgian adult population
Goscinny S, et al.
Chemosphere, 84(3), 279-288 (2011)
Determinations of hexabromocyclododecane (HBCD) isomers in channel catfish, crayfish, hen eggs and fish feeds from China by isotopic dilution LC-MS/MS
Hu X, et al.
Chemosphere, 82(5), 698-707 (2011)
Determination of hexabromocyclododecane diastereoisomers in air and soil by liquid chromatography-electrospray tandem mass spectrometry
Yu Z, et al.
Journal of Chromatography A, 1190(1-2), 74-79 (2008)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service