Skip to Content
Merck
All Photos(1)

Key Documents

01713

Sigma-Aldrich

Cryolite

synthetic, ≥97.0% (from F)

Synonym(s):

Kryolith, Sodium hexafluoroaluminate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na3AlF6
CAS Number:
Molecular Weight:
209.94
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (from F)

form

powder

quality

synthetic

reaction suitability

core: aluminum
reagent type: catalyst

impurities

≤0.5% silicic acid (as SiO2)

loss

≤0.5% loss on drying, 105 °C

cation traces

Fe: ≤500 mg/kg

storage temp.

2-30°C

SMILES string

[Na+].[Na+].[Na+].F[Al-3](F)(F)(F)(F)F

InChI

1S/Al.6FH.3Na/h;6*1H;;;/q+3;;;;;;;3*+1/p-6

InChI key

REHXRBDMVPYGJX-UHFFFAOYSA-H

Looking for similar products? Visit Product Comparison Guide

Application

Cryolite is commonly used as an electrolyte for aluminum electrolysis. Alumina is dissolved in molten cryolite is used to dissolve alumina during aluminium processing.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Chronic 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solubility of aluminum in cryolite-based melts.
Danielik V, et al.
Metallurgical and Materials Transactions A: Physical Metallurgy and Materials Science, 41(2), 430-436 (2010)
S Gelsomino et al.
The Journal of heart valve disease, 10(5), 603-610 (2001-10-18)
Left ventricular (LV) hypertrophy has been shown adversely to affect LV function and late outcome after aortic valve replacement (AVR). The study aim was to assess the time course of LV mass regression (LVMR) after AVR with a CryoLife-O'Brien stentless
P Grandjean et al.
American journal of epidemiology, 121(1), 57-64 (1985-01-01)
A cohort of 431 male cryolite workers employed for at least six months between 1924 and 1961 was identified from personnel records at the Copenhagen cryolite factory. During this period, heavy fluoride exposure resulted in at least 74 cases of
G A Valova et al.
TSitologiia i genetika, 28(3), 56-59 (1994-05-01)
Dermatoglyphic investigations which prove the existence of genetic predisposition to occupational fluorosis in workers of aluminum and criolite plants were carried out. Mathematical methods of pattern recognition were used for the multifactorial analysis. It was found that the complex analysis
T D Grekhova et al.
Meditsina truda i promyshlennaia ekologiia, (8)(8), 20-23 (1994-01-01)
Efficiency of glutamic acid for therapy of early signs of occupational fluorosis was studied in workers engaged into cryolite production. The study proved that use of glutamic acid in occupational conditions prevents progressing of metabolic disorders. The results encourage recommendations

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service