Skip to Content
Merck
All Photos(1)

Key Documents

00530580

Sennoside B

primary reference standard

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C42H38O20
CAS Number:
Molecular Weight:
862.74
Beilstein:
5723747
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

grade

primary reference standard

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

HWI

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

SMILES string

OC[C@H]1O[C@@H](Oc2cccc3[C@@H]([C@H]4c5cccc(O[C@H]6O[C@H](CO)[C@@H](O)[C@H](O)[C@H]6O)c5C(=O)c7c(O)cc(cc47)C(O)=O)c8cc(cc(O)c8C(=O)c23)C(O)=O)[C@H](O)[C@@H](O)[C@@H]1O

InChI

1S/C42H38O20/c43-11-23-31(47)35(51)37(53)41(61-23)59-21-5-1-3-15-25(17-7-13(39(55)56)9-19(45)27(17)33(49)29(15)21)26-16-4-2-6-22(60-42-38(54)36(52)32(48)24(12-44)62-42)30(16)34(50)28-18(26)8-14(40(57)58)10-20(28)46/h1-10,23-26,31-32,35-38,41-48,51-54H,11-12H2,(H,55,56)(H,57,58)/t23-,24-,25-,26+,31-,32-,35+,36+,37-,38-,41-,42?/m1/s1

InChI key

IPQVTOJGNYVQEO-OVYIKYHGSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Produced and qualified by HWI pharma services GmbH.
Exact content by quantitative NMR can be found on the certificate.

Application

Reference standard in the analysis of herbal medicinal products.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Acute hemorrhagic colitis following administration of sennosides for colon cleansing.
Burhan Ozdil et al.
The Annals of pharmacotherapy, 44(4), 770-771 (2010-03-25)
Manuk N Manukyan et al.
Surgical laparoscopy, endoscopy & percutaneous techniques, 21(2), 90-93 (2011-04-08)
The aim of this study is to assess 2 different bowel-cleansing agents. The patients were prospectively randomized to 2 arms of sodium phosphate versus Sennoside A+B calcium preparation. Laboratory assessment, body weight, height, and vital signs were obtained at baseline
Constipation and aspiration pneumonia.
Maki Takahashi et al.
Geriatrics & gerontology international, 12(3), 570-571 (2012-06-21)
Emi Matsui et al.
Biological & pharmaceutical bulletin, 34(9), 1438-1442 (2011-09-02)
Daiokanzoto (DKT), a Kampo medicine that includes the combination of two crude drugs (rhubarb and glycyrrhiza), is clinically effective for constipation. The aim of this study is to clarify the influence of glycyrrhiza, three glycyrrhiza constituents (glycyrrhizin, liquiritin, and liquiritin
Jing Xiao et al.
Wei sheng yan jiu = Journal of hygiene research, 40(3), 355-357 (2011-06-24)
To develop an analytical method for determination of sennoside A and sennoside B simultaneously in health food by high performance liquid chromatography (HPLC). Samples were extracted by ultrasound extraction and determined by HPLC with a UV detector. Using a Synergi

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service