Skip to Content
Merck
All Photos(1)

Documents

00160

Sigma-Aldrich

Acetamide

≥99.0% (GC)

Synonym(s):

Amide C2

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONH2
CAS Number:
Molecular Weight:
59.07
Beilstein:
1071207
EC Number:
MDL number:
UNSPSC Code:
12352111
eCl@ss:
39031237
PubChem Substance ID:
NACRES:
NA.77

vapor pressure

1 mmHg ( 65 °C)

Quality Level

Assay

≥99.0% (GC)

form

crystals

bp

221 °C (lit.)

mp

78-80 °C (lit.)
78-82 °C

solubility

H2O: soluble 1 gm in 0.5 ml
alcohol: soluble 1 gm in 2ml
pyridine: soluble 1 gm in 6 ml
chloroform: soluble
glycerol: soluble

SMILES string

CC(N)=O

InChI

1S/C2H5NO/c1-2(3)4/h1H3,(H2,3,4)

InChI key

DLFVBJFMPXGRIB-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Acetamide was used as a supplement for growth media and for complementation of tlyA transposon mutants.The product was used as a component for cryoprotectant solution to study the ultrastructural changes in bovine oocytes by using vitrification.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

E Fuku et al.
Cryobiology, 32(2), 139-156 (1995-04-01)
Oocytes recovered from abattoir-derived ovaries were exposed to a cryoprotectant solution (DAP213: 2 M DMSO, 1 M acetamide, 3 M propanediol, and 10% fetal calf serum in tissue culture medium 199) for less than 20 s and vitrified either at
Courtney E Maus et al.
Antimicrobial agents and chemotherapy, 49(2), 571-577 (2005-01-28)
Capreomycin, an important drug for the treatment of multidrug-resistant tuberculosis, is a macrocyclic peptide antibiotic produced by Saccharothrix mutabolis subspecies capreolus. The basis of resistance to this drug was investigated by isolating and characterizing capreomycin-resistant strains of Mycobacterium smegmatis and
A E Alves et al.
Reproduction in domestic animals = Zuchthygiene, 47(6), 1003-1008 (2012-03-06)
This study was undertaken to compare cryotolerance, in terms of viability and resumption of meiosis after warming and culture (24 and 48 h), of ex situ (isolated) and in situ (enclosed in the ovarian tissue) feline cumulus-oocyte complexes (COCs) vitrified
Cristian Varela et al.
Chemistry & biology, 19(4), 498-506 (2012-04-24)
Mycolic acids are vital components of the cell wall of the tubercle bacillus Mycobacterium tuberculosis and are required for viability and virulence. While mycolic acid biosynthesis is studied extensively, components involved in mycolate transport remain unidentified. We investigated the role
L Wang et al.
The journal of physical chemistry. B, 115(13), 3713-3724 (2011-03-17)
Infrared (IR) spectroscopy of the amide I band has been widely utilized for the analysis of peptides and proteins. Theoretical modeling of IR spectra of proteins requires an accurate and efficient description of the amide I frequencies. In this paper

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service