Skip to Content
Merck

E64-RO

Roche

E-64

N-[N-(L-3-Trans-carboxirane-2-carbonyl)-L-leucyl]-agmatine

Synonym(s):

E-64, n-[n-( l-transepoxysuccinic acid)-l-leucyl]-agmatine, n-[n-(l-3-trans-carboxirane-2-carbonyl)-l-leucyl]-4-aminobutylguanidine, n-[n-(l-3-trans-carboxirane-2-carbonyl)-l-leucyl]-agmatine.
note: another name for l-3-trans-carboxyoxiran-2-carboxylic acid acid is l-transepoxysuccinic acid. agmatine is 4-aminobutylguanidine., trans-Epoxysuccinyl-L-leucylamido(4-guanidino)butane, L-trans-3-Carboxyoxiran-2-carbonyl-L-leucylagmatine, N-(trans-Epoxysuccinyl)-L-leucine 4-guanidinobutylamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C15H27N5O5
CAS Number:
Molecular Weight:
357.41
PubChem Substance ID:
UNSPSC Code:
12352202
Beilstein/REAXYS Number:
1405664
MDL number:
NACRES:
NA.54
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

>95% (HPLC)

form

powder

mol wt

Mr 357.4

packaging

pkg of 10 mg (10874523001), pkg of 25 mg (11585681001)

manufacturer/tradename

Roche

pH range

2-10

solubility

ethanol: water (1:1): soluble 20 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)C[C@H](NC(=O)[C@@H]1O[C@H]1C(O)=O)C(=O)NCCCCNC(N)=N

InChI

1S/C15H27N5O5/c1-8(2)7-9(20-13(22)10-11(25-10)14(23)24)12(21)18-5-3-4-6-19-15(16)17/h8-11H,3-7H2,1-2H3,(H,18,21)(H,20,22)(H,23,24)(H4,16,17,19)/t9-,10+,11+/m0/s1

InChI key

LTLYEAJONXGNFG-HBNTYKKESA-N

Application

E-64 is an effective ligand for affinity purification of cysteine proteases. When coupled to a thiolated affinity matrix, binding is no longer irreversible, but specificity is retained.
E-64 is used for the inhibition of papain and other cysteine proteases such as cathepsin B and L, bromelain, and ficin during isolation and purification of proteins and enzymes.
It has been used in the affinity chromatography for the purification of hemoglobin receptors.
The inhibition of thiol proteases by E-64 appears to be of a non-competitive nature between the SH components. The inhibition is also irreversible, and is altered after gel filtration (Sephadex column) or dialysis after incubation of papain with E-64. The enzyme and inhibitor combine in an equimolar ratio.

Biochem/physiol Actions

E-64 is an irreversible, potent, and highly selective cysteine protease inhibitor. E-64 does not react with the functional thiol group of non-protease enzymes, such as L-lactate dehydrogenase or creatine kinase. E-64 will not inhibit serine proteases (except trypsin) like other cysteine protease inhibitors, leupeptin and antipain. The trans-epoxysuccinyl group (active moiety) of E-64 irreversibly binds to an active thiol group in many cysteine proteases, such as papain, actinidase, and cathepsins B, H, and L to form a thioether linkage. E-64 is a very useful cysteine protease inhibitor for use in in vivo studies because it has a specific inhibition, it is permeable in cells and tissues and has low toxicity.
Effective irreversible inhibitor of cysteine proteases that does not affect cysteine residues in other enzymes or react with low molecular weight thiols such as calpain and cathepsin B. Effective concentration 1-10 μM.
The thiol protease inhibitor E-64 specifically inhibits papain and other cysteine proteases such as cathepsin B and L, bromelain, and ficin. Cathepsin A and D are not inhibited. L-lactate dehydrogenase from porcine heart is not inhibited, although the enzyme contains a functional thiol group.

Preparation Note

Working concentration: 0.5 to 1.0 μg/ml (1.4 to 28.0 μM)
Working solution: Soluble to 20 mg/ml (stock solution) in a 1:1 (v/v) mixture of ethanol and water (vortexing or slight warming in water bath (40 °C) may facilitate dissolution). Also soluble in a neutral water/methanol solution, in water, methanol, acetic acid, pyridine and DMSO. Sparingly soluble in ethanol and propanol. Insoluble in acetone, chloroform, ethyl ether and benzene.
Storage conditions (working solution): -15 to -25 °C
Solutions are stable for one month when stored in aliquots at -15 to -25 °C.
E-64 is soluble in water. A 20 mg/ml solution can be prepared in water (heat may be needed). A suggested stock solution is a 1 mM aqueous solution. The effective concentration for use as a protease inhibitor is 1 to10 μM. Aqueous stock solutions are stable for months at -20 °C. Diluted solutions are stable for days at neutral pH. E-64 is stable from pH 2-10, but is unstable in ammonia or in HCl. E-64 is also soluble in DMSO, a 10 mM solution can be prepared in dry DMSO and stored at -20 °C. Subsequent dilutions were in culture medium. Solutions for injection were prepared by dissolving E-64 in 0.9% sodium chloride or in a minimum amount of saturated sodium bicarbonate followed by dilution with 0.9% sodium chloride (after adjusting the pH to 7.0 with acetic acid).
Soluble to 20 mg/ml in a 1:1 (v/v) mixture of ethanol and water. Solutions are stable for one month when stored in aliquots at -15 to -25 °C.

Other Notes

For life science research only. Not for use in diagnostic procedures.


Still not finding the right product?

Explore all of our products under E-64


pictograms

Health hazard

signalword

Warning

hcodes

Hazard Classifications

STOT SE 2

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

does not flash

flash_point_c

does not flash



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library