Skip to Content
Merck

DNSTP-RO

Roche

Deoxynucleoside Triphosphate Set

≤1.5% (dNDP, HPLC), ≥=98% (dNTP), ≤0.5% (dNMP, HPLC)

Synonym(s):

datp, dctp, dgtp, dttp, set of datp, dctp, dgtp, dttp

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

UNSPSC Code:
41106300
NACRES:
NA.54
Assay:
≥98% (dntp)
Form:
liquid
Solubility:
water: miscible
Storage temp.:
−20°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

description

Lithium salt

assay

≥98% (dntp)

form

liquid

composition

dNDP, ≤1.5% HPLC , dNMP, ≤0.5% HPLC , dNTP, ≥98%

packaging

pkg of 4 × 100 μL (11277049001 [4x10 μmol]), pkg of 40 × 100 μL (11922505001 [40x100mM])

manufacturer/tradename

Roche

color

colorless

solubility

water: miscible

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Deoxynucleoside Triphosphate Set consists of four separate vials of dATP, dCTP, dGTP, and dTTP. Each vial contains a 100 mM clear, colorless solution of the lithium salt (pH 7).

Application

  • Absorbance determination: A250/A260, A280/A260, A290/A260
  • DNase activity negative, according to the current Quality Control procedures
  • Suitability for PCR
Use the Deoxynucleoside Triphosphate Set, lithium salt, for PCR, RT-PCR, DNA sequencing/cycle sequencing, labeling, Reverse Transcription and all types of amplification reactions or primer-extension reactions.

Packaging

1 set containing 4 components

Preparation Note

Activator: Mg2+
Working solution: If required, the solutions can be diluted with distilled autoclaved water.

Other Notes

For life science research only. Not for use in diagnostic procedures.
The complete set of deoxynucleotides is also available in a PCR Grade. These PCR Grade deoxynucloeotides are even purer than the lithium salts, to ensure excellent performance in your PCRs. PCR Grade deoxynucleotides contain the sodium salt of each nucleotide in PCR Grade Water.

Kit Components Only

Product No.
Description

  • dATP, lithium salt 100 mM

  • dCTP, lithium salt 100 mM

  • dGTP, lithium salt 100 mM

  • dTTP, lithium salt 100 mM

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

does not flash

flash_point_c

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Global Trade Item Number

SKUGTIN
1192250500104061838708403
1127704900104061838700391

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service