Skip to Content
Merck
All Photos(8)

Key Documents

11796828001

Roche

High Pure PCR Template Preparation Kit

pkg of 100 purifications, suitable for DNA extraction

Synonym(s):

DNA extraction

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105500

usage

sufficient for 100 purifications

Quality Level

manufacturer/tradename

Roche

packaging

pkg of 100 purifications

technique(s)

DNA extraction: suitable

Related Categories

General description

Low to medium throughput genomic DNA isolation.

High Pure PCR Template Preparation Kit; Instructions For Use

Nucleic acids bind to the surface of the glass fiber fleece in the presence of a chaotropic salt (guanidine HCl). This allows the High Pure filter tube to specifically immobilize nucleic acids (both DNA and RNA) while they are freed of contaminants.

Note: A special Inhibitor Removal Buffer is included which allows the use of heparinized sample material (100 U/mL of heparin). This buffer increases the sensitivity and reproducibility of assays performed with the isolated nucleic acid, even when the sample contains heparin.

Capacity: The High Pure Spin Filter Tubes hold up to 700 μL sample volume.

Application

High Pure PCR Template Preparation Kit has been used in the extraction of genomic DNA from whole blood samples, cervical lesion specimens, gastric muscosal tissue and cervical carcinoma cell lines.
The High Pure PCR Template Preparation Kit purifies nucleic acids from a wide variety of sample materials including whole blood, cultured cells, and tissue. The isolated nucleic acids can be used for:
  • Long-template PCR
  • Real-time, quantitative PCR
  • SNP detection
  • Southern blotting
  • Cloning

Features and Benefits

The High Pure PCR Template Preparation Kit purifies nucleic acids from a wide variety of sample materials, including whole blood, cultured cells, and tissue samples. Bacteria and yeast require a specific pre-lysis treatment with lysozyme or lyticase.
  • Minimize DNA loss using a kit that removes contaminants without precipitation or other handling steps that degrade DNA.
  • Recover high molecular weight DNA (30 to 50 kb) that is suitable for long-template PCR.
  • Improve reliability and reproducibility in downstream applications (real-time, quantitative PCR).
  • Save time and maximize flexibility by preparing multiple PCR templates simultaneously.
  • Eliminate the use of hazardous organic compounds such as cesium chloride, phenol, chloroform, and ethidium bromide.

Components

  • Tissue Lysis Buffer
  • Binding Buffer
  • Proteinase K, recombinant PCR grade
  • Inhibitor Removal Buffer
  • Washing Buffer
  • Elution Buffer
  • High Pure Filter Tubes
  • Collection Tubes

Quality

DNA is isolated from 25 mg of calf thymus, 1 x 106 K562 cells, and 200 μl of EDTA whole blood. Yield is measured via OD for tissue and cell samples. The quality of the nucleic acid is controlled in an Expand Long Range PCR with a 9.3 kb amplification product for DNA derived from cells. PCR on a LightCycler® Instrument is performed on human blood research samples using kits for Factor V Leiden and CycA.

Principle

Blood cells or tissue are lysed by a short incubation with a special Lysis Buffer and Proteinase K in the presence of a chaotropic salt such as guanidine-HCl, which immediately inactivates all nucleases. Cellular nucleic acids bind selectively to special glass fibers pre-packed in the High Pure Purification Filter Tube. Bound nucleic acid is purified in a series of rapid wash-and-spin steps to remove contaminating cellular components. Finally, low salt elution releases the nucleic acid from the glass fiber. This simple method eliminates the need for organic solvent extractions and DNA precipitation, allowing for rapid purification of many samples simultaneously.

Legal Information

LightCycler is a registered trademark of Roche

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Irrit. 2 Inhalation - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High expression of ezrin predicts poor prognosis in uterine cervical cancer
Kong J, et al.
BMC Cancer, 13(1), 520-520 (2013)
Methylation-mediated transcriptional repression of microRNAs during cervical carcinogenesis
Wilting SM, et al.
Epigenetics, 8(2), 220-228 (2013)
Serum homocysteine, vitamin B12, folic acid levels and methylenetetrahydrofolate reductase (MTHFR) gene polymorphism in vitiligo
Yasar A, et al.
Disease Markers, 33(2), 85-89 (2012)
Rahmi Aslan et al.
Hepato-gastroenterology, 60(123), 601-604 (2012-10-31)
Our purpose in this study was to analyze telomere length and telomerase activity before and after eradication treatment in gastric mucosa in patients positive for H. pylori. There were two groups: a control group (n=17) and a study group (n=21).
Cielo M León et al.
Frontiers in microbiology, 8, 1907-1907 (2017-10-20)
Leishmaniasis comprises a spectrum of parasitic diseases caused by protozoans of the genus Leishmania. Molecular tools have been widely employed for the detection of Leishmania due to its high sensitivity and specificity. However, the analytical performance of molecular platforms as

Articles

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Identify causes and remedies for SDS-PAGE sample preparation challenges and optimize electrophoresis conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service