Skip to Content
Merck
All Photos(1)

Key Documents

11644793001

Roche

Cytotoxicity Detection Kit (LDH)

suitable for protein quantification, suitable for cell analysis, detection, sufficient for ≤2,000 tests

Synonym(s):

LDH

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

usage

sufficient for ≤2,000 tests

Quality Level

packaging

pkg of 1 kit

manufacturer/tradename

Roche

technique(s)

protein quantification: suitable

λmax

490 nm

application(s)

cell analysis
detection

detection method

colorimetric

storage temp.

−20°C

General description

A nonradioactive alternative to the [51Cr] and [3H]-thymidine release assay.
Colorimetric assay for the quantification of cell death and cell lysis, based on the measurement of lactate dehydrogenase (LDH) activity released from the cytosol of damaged cells into the supernatant.

Application

The Cytotoxicity Detection Kit (LDH) is a fast and simple method to quantify cytotoxicity/cytolysis based on the measurement of LDH activity released from damaged cells using the 96-well plate format. Thus, the kit can be used in many different in vitro cell systems when damage to the plasma membrane occurs. For example:
  • Detection and quantification of cell-mediated cytotoxicity
  • Determination of mediator-induced cytolysis
  • Determination of the cytotoxic potential of compounds in environmental and medical research, and in the food, cosmetic, and pharmaceutical industries
  • Determination of cell death in bioreactors
  • Testing of biocompatibility of various scaffolds, employed in bone tissue engineering, for bone cell growth

Packaging

1 kit containing 2 components.

Preparation Note

Working solution: Vial 1 Catalyst:
Reconstitute the lyophilisate in 1 ml double dist. water for 10 minutes and mix thoroughly.

Vial 2 Dye solution:
Ready-to-use solution.

Reaction mixture
For 100 tests: Shortly before use, mix 250 μl of vial 1 with 11.25 ml of vial 2.
For 400 tests: Shortly before use, add the total volume of catalyst (1 ml) to the total volume
of dye solution (45 ml) and mix well.

Note: The Reaction mixture should not be stored; prepare immediately before use.
Storage conditions (working solution): 2 to 8 °C
The lyophilizate is stable at 2 to 8 °C.
The reconstituted catalyst solution is stable for four weeks when stored at 2 to 8 °C.
Once thawed, the dye solution is stable for several weeks when stored at 2 to 8 °C.

Kit Components Only

Product No.
Description

  • Catalyst (Diaphorase/NAD+ mixture)

  • Dye Solution (INT and sodium lactate)

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sabine Sewing et al.
PloS one, 11(7), e0159431-e0159431 (2016-07-22)
Single stranded oligonucleotides (SSO) represent a novel therapeutic modality that opens new space to address previously undruggable targets. In spite of their proven efficacy, the development of promising SSO drug candidates has been limited by reported cases of SSO-associated hepatotoxicity.
Guo-qian He et al.
Molecular brain, 8, 88-88 (2015-12-25)
Growth arrest and DNA-damage inducible protein 45 beta (Gadd45b) is serving as a neuronal activity sensor. Brain ischemia induces the expression of Gadd45b, which stimulates recovery after stroke and may play a protective role in cerebral ischemia. However, little is
Kacper A Wojtal et al.
American journal of physiology. Gastrointestinal and liver physiology, 307(7), G673-G688 (2014-06-28)
Solute carrier (SLC) transporters mediate the uptake of biologically active compounds in the intestine. Reduced oxygenation (hypoxia) is an important factor influencing intestinal homeostasis. The aim of this study was to investigate the pathophysiological consequences of hypoxia on the expression
M C Skinner et al.
Antimicrobial agents and chemotherapy, 54(2), 627-636 (2009-12-17)
Topical microbicides for prevention of sexually transmitted diseases (STDs) would be especially useful for women who are not able to persuade their partner(s) to take precautions. Many topical microbicides are in various stages of development, based on a variety of
Shilpee Singh et al.
Nitric oxide : biology and chemistry, 32, 21-28 (2013-04-17)
Surges of nitric oxide compromise mitochondrial respiration primarily by competitive inhibition of oxygen binding to cytochrome c oxidase (complex IV) and are particularly injurious in neurons, which rely on oxidative phosphorylation for all their energy needs. Here, we show that

Articles

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Quality control guidelines to maintain high quality authenticated and contamination-free cell cultures. Free ECACC handbook download.

Related Content

Cytotoxicity Detection Kit (LDH) Protocol & Troubleshooting

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service