Skip to Content
Merck
All Photos(1)

Key Documents

11333062910

Roche

Anti-Digoxigenin

from mouse IgG1κ (clone 1.71.256)

Synonym(s):

anti-digoxigenin, digoxigenin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

clone 1.71.256, monoclonal

form

lyophilized

packaging

pkg of 100 μg

manufacturer/tradename

Roche

technique(s)

ELISA: 2-4 μg/mL
immunocytochemistry: 0.5-2 μg/mL
immunohistochemistry: 0.5-2 μg/mL
western blot: 0.5-2 μg/mL

isotype

IgG1κ

storage temp.

2-8°C

General description

Contents: Lyophilizate
Digoxigenin is a hapten, useful in labeling nucleic acids and in detection systems. Probes labeled with digoxigenin has greater sensitivity equivalent to that of radioactive probes, allows faster detection. It is less hazardous and has increased shelf life.

Specificity

The monoclonal antibody reacts with free and bound digoxigenin.

Application

Use Anti-Digoxigenin antibody for the detection of digoxigenin-labeled componds using:
  • ELISA
  • Immunohistocytochemistry
  • In situ hybridization
  • Western blot
  • Immunofluorescence staining
  • FISH (fluorescent in situ hybridization)

Biochem/physiol Actions

In the presence of Na+, Mg2+ and adenosine triphosphate (ATP), digoxigenin inhibits sodium pumps.

Features and Benefits

  • Specific to free and bound digoxigenin
  • The antibody is not stabilized with protein and therefore suitable as coating antibody in Sandwich-ELISAs and for labeling procedures. For example it can be conjugated with enzymes or fluorescence dyes for direct detection of digoxigenin-labeled samples.

Preparation Note

Working concentration: Working concentration of antibody depends on application and substrate. The following concentrations should be taken as a guideline:
  • ELISA: for coating: 2 to 4 μg/ml
  • Immunohistocytochemistry: 0.5 to 2 μg/ml
  • In situ hybridization: 0.2 to 0.4 μg/ml
  • Western blot: 0.5 to 2 μg/ml

Working solution: For coating applications
phosphate buffered saline, pH 7.4

Reconstitution

Add 1 ml double-distilled water to a final concentration of 100 μg/ml.

Storage and Stability

The lyophilized antibody is stable at +2 to +8°C. The reconstituted antibody solution is stable up to 6 months at +2 to +8°C. The solution can be stored in aliquots at -15 to -25°C. Avoid repeated freezing and thawing.

Analysis Note

There are no cross reactivities known.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ilyas Chachoua et al.
Nature communications, 13(1), 204-204 (2022-01-13)
Abnormal WNT signaling increases MYC expression in colon cancer cells in part via oncogenic super-enhancer-(OSE)-mediated gating of the active MYC to the nuclear pore in a poorly understood process. We show here that the principal tenet of the WNT-regulated MYC
Carla Winterling et al.
RNA biology, 11(1), 66-75 (2014-01-21)
A growing body of evidence suggests the non-protein coding human genome is of vital importance for human cell function. Besides small RNAs, the diverse class of long non-coding RNAs (lncRNAs) recently came into focus. However, their relevance for infection, a
Affinity Labeling of a Sulfhydryl Group in the Cardiacglycoside Receptor Site of Na+/K+-ATPase by N-Hydroxysuccinimidyl Derivatives of Digoxigenin
<BIG>Antolovic R, et al.</BIG>
European Journal of Biochemistry, 227, 61-67 (1995)
Nonradioactive labeling of probe with digoxigenin by polymerase chain reaction
<BIG>Lion T and Haas OA</BIG>
Analytical Chemistry, 188, 335-337 (1990)
Kristiina Joensuu et al.
Breast cancer : basic and clinical research, 7, 23-34 (2013-03-22)
Breast cancer can recur even decades after the primary therapy. Markers are needed to predict cancer progression and the risk of late recurrence. The estrogen receptor (ER), progesterone receptor (PR), human epidermal growth factor receptor-2 (HER2), proliferation marker Ki-67, and

Articles

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Digoxigenin (DIG) labeling methods and kits for DNA and RNA DIG probes, random primed DNA labeling, nick translation labeling, 5’ and 3’ oligonucleotide end-labeling.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service