Yes, the Millex High Output and Chemical Duty pumps are CE approved.
WP6211560
High Output Pump, 115 V/60 Hz
Synonym(s):
Laboratory vacuum pump
About This Item
Recommended Products
AC/DC input
115 V, 60 Hz
shipped in
ambient
General description
The High Output Pump features a piston-driven design for greater power.
The Chemical Duty Pump has a chemically-resistant head and diaphragm for use with corrosive chemicals and solvents.
Both pumps come with 70 cm of 1/4" tubing and a Millex FA50 filter for in-line moisture protection. Both are UL listed and CE marked.
The High Output and Chemical Duty pumps can support high flow rates to decrease process filtration time. The High Output Pump features a piston-driven design for greater power. The Chemical Duty Pump has a chemically-resistant head and diaphragm for use with corrosive chemicals and solvents. Both pumps come with 70 cm of 1/4 in. tubing and a Millex™-FA50 filter for in-line moisture protection. Both are UL-listed and CE-marked.
Features & Benefits
- High flow rates decrease filtration process time
- Optional chemical-resistant internal components for compatibility with corrosive vapors
- Portable design allows for easy sharing between workstations
Applications
Vacuum Source for Filter Holders, Manifolds, and Devices, including Samplicity Filtration System, SNAP i.d. 2.0 System, MultiscreenHTS Filtration Manifold, and Stericup Filters
Principle
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
An overview of microplastics, testing regulations, and filter selection for microplastic sample collection, preparation, and analysis.
-
Is product WP6211560 CE Approved?
1 answer-
Helpful?
-
-
HI is the WP6211560 CE Approved?
1 answer-
Yes, the Millex High Output and Chemical Duty pumps are CE approved.
Helpful?
-
-
Is the high output pump Cat No WP6211560, CSA approved
1 answer-
No, this pump is not CSA approved.
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service