VL-1L-BK
Intermediates For Further Manufacturing Use (FFMU) - Rare Specificities
Anti-Fya from the clone P3TIM, Human IgG Antibody, Intermediate for further manufacturing use
Synonym(s):
Anti-Fya (clone P3TIM), FFMU
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41116102
NACRES:
NA.41
Recommended Products
biological source
human
Quality Level
antibody form
unpurified
clone
P3TIM, monoclonal
form
liquid
packaging
pkg of 1 L
concentration
5-15 mg/mL protein (Total Protein Range)
technique(s)
agglutination assay: suitable
pH
6.7-7.7
isotype
IgG
shipped in
ambient
storage temp.
2-8°C
Related Categories
General description
Standardized, concentrated cell culture supernatant which has been diafiltered with phosphate buffered saline.; For the formulation of Anti-Fya blood typing reagents or devices.; Contains 0.1% (w/v) Sodium Azide; Shelf Life:21 months from date of filtration.
Application
Antibody intermediate for further manufacturing use
Production of an in vitro blood typing product or device
Legal Information
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service