Skip to Content

System maintenance is scheduled for 11 Sep, 08:00 PM - 12 Sep, 00:00 AM CST [12 Sep 03:00 AM to 07:00 AM CET] and will affect some web and offline transactions, including quotes, price checks, and order status inquiries. We apologize for any inconvenience.

Merck

Skip To

SEGPU1145

Stericup® E-GP Funnel-less Sterile Vacuum Filtration System

pore size 0.22 μm, polyethersulfone membrane, filter capacity 1000 mL, 45 mm thread, pack of 12 ea

greener alternative

Greener Alternative

Synonym(s):

Stericup® E-GP Eco-friendly Filter Unit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NB.24
UNSPSC Code:
41104914
Pore size:
0.22 μm pore size
Filter capacity:
1000 mL
Packaging:
pack of 12 ea
Material:
polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

material

polyethersulfone membrane, polystyrene bottle, polystyrene funnel(s), polystyrene housing

product line

Stericup®

feature

hydrophilic

packaging

pack of 12 ea

greener alternative product characteristics

Waste Prevention
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

parameter

max. temp. at 45 °C

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

220 mm

diam.

11.3 cm

filter capacity

1000 mL

filter diam.

73 mm

filtration area

40 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet thread(s), 45 mm thread

greener alternative category

shipped in

ambient

storage temp.

no temp limit

General description

We are committed to bringing you Greener Alternative Products, which belong to one of the four categories of greener alternatives . This product is a Design for Sustainability (DfS) developed product. Click here to view its DfS scorecard.
Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

The Stericup® E and Steritop® E sterile filters are designed as a Greener Alternative for vacuum sterile filtration that significantly reduces plastic and packaging.

This Stericup® E is a 45 mm thread collar which uses a 0.22 μm pore size polyethersulfone (PES) membrane filter unit combined with a 1000 mL receiver flask and cap for processing and storage. The 45 mm thread directly connects to wider neck media bottles (such as Gibco® or glass bottles).

Use only glass or plastic bottles designed for vacuum applications.

Application

Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions
For Research Use Only

Features and Benefits

  • The filter collar disconnects from the receiver bottle with just a quarter turn—reducing the likelihood of spillage and making it easier to manipulate on the bench or under the hood.
  • Bold product identification on the filter collar plainly displays membrane characteristics, including composition and pore size, for rapid, application-appropriate product selection.
  • Click seal cap is flanged and modified to enable a firm grip in wet or dry conditions—and a tactile stop confirms secure closure. Worry less about the possibility of spilling or contamination of sterilized contents.
  • Frosted, ample writing surface on the receiver bottle and lighter cap color facilitate clear labelling to improve legibility and reduce unreadable smears.
  • Click seal cap rests on the side to avoid risk of contamination.
  • Receiver bottles are stackable to save storage space.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
STERITOP is a registered trademark of Merck KGaA, Darmstadt, Germany
Stericup is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SEGPU0538SEGPT0038S2GVU02RE
description

pore size 0.22 μm, polyethersulfone membrane, filter capacity 1000 mL, 45 mm thread, pack of 12 ea

description

pore size 0.22 μm, polyethersulfone membrane, filter capacity 500 mL, 38 mm (1 1/2 in.) thread, pack of 12 ea

description

pore size 0.22 μm, polyethersulfone membrane, filter capacity 150 mL × 1000 mL, 38 mm (1 1/2 in.) thread, pack of 12 ea, Greener Alternative

description

pore size 0.22 μm, PVDF membrane, filter capacity 250 mL, pack of 12 ea

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

pore size

0.22 μm pore size

filter capacity

1000 mL

filter capacity

500 mL

filter capacity

150 mL , 150 mL × 1000 mL

filter capacity

250 mL

material

polystyrene housing, polystyrene funnel(s), polyethersulfone membrane, polystyrene bottle

material

polystyrene funnel(s), polystyrene bottle, polyethersulfone membrane, polystyrene housing

material

polystyrene funnel(s), polystyrene housing, polystyrene bottle, polyethersulfone membrane

material

polystyrene bottle, polystyrene housing, polystyrene funnel(s), PVDF membrane

fitting

45 mm thread, inlet thread(s)

fitting

inlet thread(s), 38 mm (1 1/2 in.) thread

fitting

38 mm (1 1/2 in.) thread, inlet thread(s)

fitting

inlet funnel, outlet flask (Receiver)

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

feature

hydrophilic

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea

packaging

pack of 12 ea


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

1–10 of 15 Questions  
  1. What is the chemical compatibility of the various Stericup® and Steritop® filters?

    What is the chemical compatibility of the various Stericup® and Steritop® filters?

    1 answer
    1. Helpful?

  2. The User Guide says “Use only glass or plastic bottles designed for vacuum applications”. Can you specify which bottles are compatible?

    1 answer
    1. Steritop® Quick Release bottle top units require a vacuum-rated receiver bottle with either a GL33 or GL45 mm closure thread as specified. Steritop® E bottle top units require a vacuum-rated receiver bottle with a GL45 mm closure thread. Bottles no larger than 2 L should be used.
      Glass bottles, GL33:
      • Wheaton media bottles (glass), 500 mL (Product Z364886)
      • Wheaton media bottles (glass), 1 L (Product Z364894)
      Glass bottles, GL45
      • Corning® glass media storage bottle, 125 mL (Product CLS1399125)
      • Corning® glass media storage bottle, 250 mL (Product CLS1399250)
      • Corning® glass media storage bottle, 500 mL (Product CLS1399500)
      • Corning® glass media storage bottle, 1 L (Product CLS13991L)
      • Pyrex® round media storage bottle, 500 mL (Product CLS1395500)
      • Pyrex® round media storage bottle, 1 L (Product CLS13951L)
      • KIMBLE® KIMAX® Media bottle, 250 mL (Product DWK14395250)
      • KIMBLE® KIMAX® Media bottle, 500 mL (Product DWK14395500)
      • KIMBLE® KIMAX® Media bottle, 1 L (Product DWK143951000)
      Plastic (pre-sterile) bottles, GL45:
      • Stericup® Quick Release Receiver flask, 250 mL (Product S200B02RE)
      • Stericup® Quick Release Receiver flask, 500 mL (Product S200B05RE)
      • Stericup® Quick Release Receiver flask, 1000 mL (Product S200B10RE)

      Helpful?

  3. What Bacterial Endotoxins Test is used?

    1 answer
    1. The test method used complies with USP 85 and EP 2.6.14 to ensure the absence of endotoxin in our products.

      Helpful?

  4. Are Stericup® and Steritop® filters GMP-certified?

    1 answer
    1. No, Stericup® and Steritop® devices are not GMP-approved.

      Helpful?

  5. What is the maximum vacuum pressure for filtration?

    1 answer
    1. 700 mm Hg (27.5” Hg, 0.9 bar) differential vacuum at 25 °C (77 °F)

      Helpful?

  6. Can we filter oil-type liquids with Stericup® filters?

    Can we filter oil-type liquids with Stericup® filters?

    1 answer
    1. Likely not, due to pore size/possible fouling/chemical compatibility. Refer to our guide, "Chemical compatibility of filter components": https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/product/documents/302/855/membrane-chemical-compatibility-ds2532en-ms.pdf

      Helpful?

  7. Can Stericup® and Steritop® filters be used to prepare solutions that will be administered to patients?

    1 answer
    1. No, all Stericup® and Steritop® devices are Research Use Only.

      Helpful?

  8. What is the maximum processing volume for Stericup® and Steritop® filters? Can I add more volume after initial filtration?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single-use application. The maximum volume is dependent on the receiver bottle capacity. Stericup® filters come with a receiver bottle with a specified maximum volume. For Steritop® filters, the maximum recommended volume is 2 L with an appropriate-sized receiver bottle. Larger processing volumes can result in membrane fouling, increase chance of spillage and contamination, and lead to possible airlock*.
      *Note that devices with PES membranes can airlock if filtered to dryness. Some liquid must be left on top of the filter, then the system stopped, receiver changed or emptied, and more sample volume added before restart. This increases the chance of spillage.

      Helpful?

  9. What is the expiration date of Stericup® and Steritop® filters?

    1 answer
    1. Typically 3 years from sterilization.

      Helpful?

  10. Can Stericup® and Steritop® filters be reused?

    1 answer
    1. All Stericup® and Steritop® filters were developed for single use, disposable processing.

      Helpful?

1–10 of 15 Questions  

Reviews

No rating value

Active Filters