Skip to Content
Merck
All Photos(1)

Key Documents

SCGPS05RE

Millipore

Millipore® Steritop® Vacuum Bottle Top Filter

pore size 0.22 μm, polyethersulfone membrane, funnel capacity 500 mL, 33 mm outlet thread(s), pack of 12 ea

Synonym(s):

Steritop Threaded Bottle Top Filter, Steritop®-GP Quick Release Bottle Top Filter, bottle top sterile filter unit, bottle top vacuum filter, bottle top vacuum filtering unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031623
NACRES:
NB.24

material

polyethersulfone membrane
polystyrene funnel(s)
polystyrene housing

Quality Level

sterility

sterile; irradiated

product line

Steritop®

feature

hydrophilic

packaging

pack of 12 ea

parameter

45 °C max. temp.

description

2.4 mL (hold-up volume)

technique(s)

sterile filtration: suitable

H

148 mm

capacity

500 mL

diam.

12.5 cm

filter diam.

73 mm

filtration area

40 cm2

funnel capacity

500 mL

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

inlet funnel
33 mm outlet thread(s)

shipped in

ambient

General description

Work with ease and filter with confidence with Stericup® and Steritop® Filtration Systems.

Stericup ® and Steritop ® sterile filtration devices use reliable, high-performance membranes for optimum sterile filtration of laboratory reagents, buffers, and media. These devices combine superior flow rates and throughput with low non-specific binding and a stable, no-tip design.

This Steritop ® 500 mL bottle top 0.22 μm pore size polyethersulfone (PES) membrane filter unit can be used on bottles with 33 mm openings, , designed for vacuum applications.

Application

For Research Use Only
Stericup® and Steritop® systems are designed to reduce foaming and protein denaturation while maximizing flow and can be used for sterile vacuum filtration of aqueous solutions such as tissue culture media and biological fluids.

  • Tissue culture media +/- additives
  • Buffers
  • Biological solutions

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
STERITOP is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.

The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.

The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.

The FPP Purospher® STAR RP-8e column offers a reliable and consistent assay with strong peak shapes and linearity, ensuring precise quantification.

Related Content

What are you filtering out? What do you want to keep? Pick the right membrane for your filtration.

What are you filtering out? What do you want to keep? Pick the right membrane for your filtration.

What are you filtering out? What do you want to keep? Pick the right membrane for your filtration.

What are you filtering out? What do you want to keep? Pick the right membrane for your filtration.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service