Skip to Content
Merck
All Photos(3)

Key Documents

SCC115

Sigma-Aldrich

CH27 Mouse B Cell Lymphoma Cell Line

Mouse

Synonym(s):

CH27 [Mouse lymphoma]

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.81

Product Name

CH27 Mouse B Cell Lymphoma Cell Line, CH27 mouse B cell lymphoma celll line is a useful model for the study of B-cell biology and tumor immunology.

biological source

mouse

Quality Level

shipped in

ambient

General description

CH27 Mouse B Cell Lymphoma Cell Line is one of the CH series of mouse B-cell lymphomas that arise from the spleen cells of B10.H-2a-H-4b p/Wts (2a4b) mice, either spontaneously or after transplantation of the spleen cells to syngeneic or F1 hybrid recipients .

CH27 cells express surface IgM and Ia antigens along with Ly-1 (CD5) and Ig which bind sheep (SRBC) and chicken erythrocytes and bromelain-treated mouse erythrocytes . The hapten recognized by SRBC-binding CH lymphomas is a phospholipid, either phosphatidylchone or sphingomyelin . Phospholipids are a common component of cell membranes and thus cells expressing immunoglobulin with specificity for SRBC can also be found in the spleens of normal mice . This suggests that CH lymphomas represent neoplastic analogues of Ly-1+ normal B-cells .

Cell Line Description

Cancer Cells

Application

Research Category
Cancer

Immune Response

Inflammation & Immunology
This product is intended for sale and sold solely for internal non-commercial research use per the terms of the “Restricted Use Agreement” as detailed in the product documentation. For information regarding any other uses, please contact licensing@emdmillipore.com.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested negative for infectious diseases by a Mouse Essential CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are verified to be of mouse origin and negative for inter-species contamination from rat, chinese hamster, Golden Syrian hamster, human and non-human primate (NHP) as assessed by a Contamination CLEAR panel by Charles River Animal Diagnostic Services.
• Cells are negative for mycoplasma contamination

Storage and Stability

Store in liquid nitrogen. The cells can be cultured for at least 10 passages after initial thawing without significantly affecting the cell marker expression and functionality.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Transplantable B-cell lymphomas in B10. H-2aH-4bp/Wts mice.
L L Lanier et al.
Immunogenetics, 16(4), 367-371 (1982-01-01)
T J Mercolino et al.
The Journal of experimental medicine, 163(1), 155-165 (1986-01-01)
Cells from 6 of 14 different Ly-1+ murine B cell lymphomas bound to synthetic liposomes encapsulating fluorescein. The liposomes were made from distearoyl phosphatidyl choline (DSPC), distearoyl phosphatidyl glycerol (DSPG), and cholesterol. In all cases, liposome binding was due to
C A Pennell et al.
Proceedings of the National Academy of Sciences of the United States of America, 82(11), 3799-3803 (1985-06-01)
A series of 27 B-cell lymphomas (designated the CH series), induced in B10.H-2aH-4b p/Wts mice by intense adoptive immunization with sheep erythrocytes, was found to represent a subset of the total B-cell repertoire. This subset was characterized by expression of
The CH series of murine B cell lymphomas: neoplastic analogues of Ly-1+ normal B cells.
G Haughton et al.
Immunological reviews, 93, 35-51 (1986-10-01)
C A Pennell et al.
Journal of immunology (Baltimore, Md. : 1950), 141(8), 2788-2796 (1988-10-15)
The majority of the characterized Ly-1+ B cell lymphomas of B10.H-2aH-4bp/Wts origin (the CH series) bear surface Ig related by Ag specificity or idiotype or both. To determine the genetic basis for these structural similarities, we have sequenced the VH

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service