Skip to Content
Merck
All Photos(3)

Key Documents

SCC064

Sigma-Aldrich

LX-2 Human Hepatic Stellate Cell Line

Human

Synonym(s):

HSC, Perisinusoidal cells, Ito cells, Hepatic lipocytes, Hepatic pericytes

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514
eCl@ss:
32011203
NACRES:
NA.75

product name

LX-2 Human Hepatic Stellate Cell Line, a highly suitable model of human hepatic fibrosis

biological source

human

Quality Level

technique(s)

cell culture | stem cell: suitable

shipped in

liquid nitrogen

General description

Hepatic stellate cells are a major cell type responsible for liver fibrosis following their activation into fibrogenic myofibroblast-like cells in diseases such as chronic alcoholism, hepatitis B and C, fatty liver disease, obesity and diabetes. There is an increasing need for renewable cell culture models that faithfully recapitulate their in vivo phenotype, particularly for human studies.

LX-2 was generated by immmortalization of primary human hepatic stellate cells with the SV40 large T antigen followed by selective culture of early passaged cells in low serum media conditions.

Immortalized LX-2 was established by Xu et al to overcome issues of culture variability and to provide a stable and unlimited source of human hepatic stellate cells that are homogeneous. These cell lines have been extensively characterized and retain key features of cytokine signaling, neuronal gene expression, retinoid metabolism, and fibrogenesis, making them highly suitable for culture based studies of human hepatic fibrosis.

Application

Research Category
Infectious Diseases

Inflammation & Immunology

Stem Cell Research

Components

1) ≥1X106 viable LX-2 Human Hepatic Stellate Cells: (Catalog No. SCC064). Store in liquid nitrogen.

Quality

• Each vial contains ≥ 1X106 viable cells.
• Cells are tested by PCR and are negative for Hepatitis A, B, C and HIV-1 & 2 viruses.
• Cells are negatrive for mycoplasma contamination.

Storage and Stability

LX-2 cells should be stored in liquid nitrogen. The cells can be passage for at least 10 passages without significantly affecting the cell marker expression and functionality. LX-2 cells have been successfully expanded past passage 50 in culture.

Other Notes

This product is intended for sale and sold solely to academic institutions for internal academic research use per the terms of the "Academic Use Agreement" as detailed in the product documentation. For information regarding any other use, please contact licensing@emdmillipore.com.

Disclaimer

RESEARCH USE ONLY. This product is regulated in France when intended to be used for scientific purposes, including for import and export activities (Article L 1211-1 paragraph 2 of the Public Health Code). The purchaser (i.e. enduser) is required to obtain an import authorization from the France Ministry of Research referred in the Article L1245-5-1 II. of Public Health Code. By ordering this product, you are confirming that you have obtained the proper import authorization.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Isolated hepatic lipocytes and Kupffer cells from normal human liver: morphological and functional characteristics in primary culture.
Friedman, S L, et al.
Hepatology, 15, 234-243 (1992)
Interleukin-21 mediates hepatitis B virus-associated liver cirrhosis by activating hepatic stellate cells.
Feng, Guohua, et al.
Hepatology Research : the Official Journal of the Japan Society of Hepatology (2013)
Ganesh Satyanarayana et al.
Theranostics, 11(19), 9331-9341 (2021-10-15)
Rationale: Fibrosis is a pathologic condition of abnormal accumulation of collagen fibrils. Collagen is a major extracellular matrix (ECM) protein synthesized and secreted by myofibroblasts, composing mainly (Gly-X-Y)n triplet repeats with >30% Gly residue. During fibrosis progression, myofibroblasts must upregulate
Ching-Fen Wu et al.
Journal of cellular biochemistry, 114(3), 541-550 (2012-09-11)
Hepatitis C virus core protein (HCVcp), which is secreted by infected cells, is reported as an immunomodulator in immune cells. However, the effects of HCVcp on hepatic stellate cells (HSCs), the key cells in liver fibrosis, still remain unclear. In
TRAIL regulates collagen production through HSF1-dependent Hsp47 expression in activated hepatic stellate cells.
Park, Soo-Jung, et al.
Cellular Signalling, 25, 1635-1643 (2013)

Related Content

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Learn how to use our cell culture tested, ready-to-use MilliShot™ single dose antibiotic solutions, conveniently packaged in one-time use vials.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service