S55535BC
Polypropylene Tubes
BioControl, For use with Gemini, size 3.5 mL
Synonym(s):
Sample tubes
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
UNSPSC Code:
41122409
NACRES:
NC.34
Recommended Products
material
polypropylene
packaging
pkg of 1000 units (Bulk packed)
manufacturer/tradename
BioControl Systems S-55.535*
Gemini®
technique(s)
ELISA: suitable
enzyme immunoassay: suitable
size
3.5 mL
application(s)
food and beverages
pathogen testing
compatibility
for use with Gemini (78000)
storage temp.
room temp
General description
The polypropylene tubes (Sample tubes) are the consumable accessories of the Gemini® system. Gemini® is a fully automated microplate processor compatible with all Enzyme immunoassays and is pre-programmed for TRANSIA® Plate assays. Polypropylene tubes have greater thermal, chemical, and mechanical stability. Material is biologically inert and is ideally suited for immunoassays and storage of samples. It offers increased capacity over other systems for high throughput testing.
Application
Polypropylene Tubes being compatible for use with Gemini® system, contribute to the automation of various procedures such as ELISA and other diagnostic Enzyme Immunoassays.
Components
Tubes, PP, 3.5 mL
Legal Information
Gemini is a registered trademark of Stratec
TRANSIA is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service