Skip to Content
Merck
All Photos(2)

Key Documents

RPTMAG-86K

Millipore

MILLIPLEX® Rat Pituitary Magnetic Bead Panel - Endocrine Multiplex Assay

The analytes available for this multiplex kit are: ACTH, BDNF, FSH, GH, LH, Prolactin, TSH.

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
eCl@ss:
32161000
NACRES:
NA.84

Quality Level

species reactivity

rat

manufacturer/tradename

Milliplex®

assay range

accuracy: 75%
(GH)

accuracy: 88%
(ACTH)

accuracy: 89%
(LH)

accuracy: 90%
(TSH)

sensitivity: 0.38 pg/mL
(BDNF)

sensitivity: 0.87 pg/mL
(TSH)

sensitivity: 1.95 pg/mL
(ACTH)

sensitivity: 3.28 pg/mL
(LH)

sensitivity: 4.64 pg/mL
(Prolactin)

sensitivity: 6.50 pg/mL
(GH)

sensitivity: 7.62 pg/mL
(FSH)

standard curve range: 16-50,000 pg/mL
(GH, Prolactin)

standard curve range: 3.2-10,000 pg/mL
(ACTH, BDNF, LH, TSH)

standard curve range: 32-100,000 pg/mL
(FSH)

inter-assay cv: <20%
intra-assay cv: <10%

technique(s)

multiplexing: suitable

detection method

fluorometric (Luminex xMAP)

shipped in

wet ice

General description

Called the “master gland” because it controls many other endocrine glands, the pituitary secretes several key hormones that play important roles in the regulation of metabolism, growth, and reproduction. Piuitary hormones are responsible for the stimulation of the adrenal gland (ACTH), thyroid gland (TSH), ovaries and testes (FSH, LH) and breast milk production (prolactin), as well as blood pressure and the amount of water excreted by the kidneys. Through its connection with the pituitary, the hypothalamus modulates endocrine function by detecting hormone levels. Brain-Derived Neurotrophic Factor (BDNF) is a member of NGF family of neurotrophic factors which are required for differentiation and survival of specific neuronal sub-populations.

MILLIPLEX® Rat Pituitary Panel is a 7-plex kit to be used for the simultaneous quantification of any or all of the following analytes in rat serum or plasma samples, rat tissue extract, or cell/tissue culture supernatant samples: Growth Hormone (GH), Thyroid-Stimulating Hormone (TSH), Adrenocorticotropic Hormone (ACTH), Luteinizing Hormone (LH), Prolactin, Follicle-Stimulating Hormone (FSH), and Brain-Derived Neurotrophic Factor (BDNF). This kit uses a 96-well format, contains a lyophilized standard cocktail, two internal assay quality controls and can measure up to 38 samples in duplicate.

The Luminex® xMAP® platform uses a magnetic bead immunoassay format for ideal speed and sensitivity to quantitate multiple analytes simultaneously, dramatically improving productivity while conserving valuable sample volume.

Panel Type: Endocrine

Application

  • Analytes: Adrenocorticotropic Hormone (ACTH), Brain-Derived Neurotrophic Factor (BDNF), Follicle-Stimulating Hormone (FSH), Growth Hormone (GH), Luteinizing Hormone (LH), Prolactin, Thyroid-Stimulating Hormone (TSH)
  • Recommended Sample Type: Rat serum, plasma, cell/tissue culture supernatants or lysates
  • Recommended Sample Dilution: 25 μL per well 1:3 diluted serum or plasma; cell/tissue culture samples may require dilution in an appropriate control medium.
  • Assay Run Time: Overnight (16-18 hours) at 2-8°C
  • Research Category: Endocrine
  • Research Subcategory: Metabolism

Features and Benefits

Design your multiplex kit by choosing available analytes within this panel.

Legal Information

Luminex is a registered trademark of Luminex Corp
MILLIPLEX is a registered trademark of Merck KGaA, Darmstadt, Germany
xMAP is a registered trademark of Luminex Corp

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vivi F H Jensen et al.
Scientific reports, 10(1), 5609-5609 (2020-03-30)
Maternal hypoglycaemia throughout gestation until gestation day (GD)20 delays foetal growth and skeletal development. While partially prevented by return to normoglycaemia after completed organogenesis (GD17), underlying mechanisms are not fully understood. Here, we investigated the pathogenesis of these changes and
Global but not gonadotrope-specific disruption of Bmal1 abolishes the luteinizing hormone surge without affecting ovulation.
Chu, A; Zhu, L; Blum, ID; Mai, O; Leliavski, A; Fahrenkrug, J; Oster, H; Boehm, U; Storch, KF
Endocrinology null
Ivraym B Barsoum et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 27(7), 2657-2666 (2013-04-10)
Testes contain two distinct Leydig cell populations during development: fetal and adult Leydig cells (FLCs and ALCs, respectively). ALCs are not derived from FLCs, and it is unknown whether these two populations share common progenitors. We discovered that hedgehog (Hh)
Mirjam Kowallick et al.
Frontiers in pediatrics, 9, 723928-723928 (2021-11-23)
Introduction: High oxygen concentrations have been identified as one factor contributing to the pathogenesis of the retinopathia of prematurity, chronic lung disease of the preterm infant and preterm brain injury. Preterm infants also show short- and long-term alterations of the
Jennifer M Rojas et al.
Toxicologic pathology, 46(7), 777-798 (2018-10-23)
The obese rodent serves as an indispensable tool for proof-of-concept efficacy and mode-of-action pharmacology studies. Yet the utility of this disease model as an adjunct to the conventional healthy animal in the nonclinical safety evaluation of anti-obesity pharmacotherapies has not

Related Content

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

MILLIPLEX® toxicity assays advance liver, kidney, and genotoxicity research by simultaneously measuring multiple toxicity biomarkers.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service