Skip to Content
Merck
All Photos(1)

Key Documents

PX1235

Supelco

Piperidine

Synonym(s):

Piperidine, Hexahydropyridine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H11N
CAS Number:
Molecular Weight:
85.15
MDL number:
UNSPSC Code:
12191504
EC Index Number:
203-813-0
bp:
106 °C/1013 hPa
vapor pressure:
34 hPa ( 20 °C)

vapor pressure

34 hPa ( 20 °C)

Quality Level

Assay

≥99.5% (GC)

form

liquid

impurities

≤0.2% Water (H2O)

color

APHA: ≤50

pH

12.6 (20 °C, 100 g/L in H2O)

bp

106 °C/1013 hPa

mp

-10.8 °C

transition temp

flash point 16 °C

density

0.86 g/cm3 at 20 °C

shipped in

ambient

storage temp.

room temp

SMILES string

N1CCCCC1

InChI

1S/C5H11N/c1-2-4-6-5-3-1/h6H,1-5H2

InChI key

NQRYJNQNLNOLGT-UHFFFAOYSA-N

Application

Piperidine can be used as:
  • A Fmoc removal agent in solid-phase peptide synthesis.
  • An organic structure-directing agent in the synthesis of ferrierite zeolites and CoAPO (cobalt aluminophosphate) materials.
  • A reagent in the preparation of N-(2,4-dinitrophenyl)piperidine by reacting with 2,4-dinitrochlorobenzene via aromatic nucleophilic substitution reaction.

Packaging

M-Bottle for Solvents
As a global leader in lab reagents, we are constantly looking for new ways to optimize the safety of our products. The newly developed 4L solvent bottle design features advanced sealing technology that eliminates leaks to make the handling of solvents safer and more convenient than ever before.
See all the new features here!

Analysis Note

Assay (GC): 99.5% min
Color (APHA): 50APHA max
Form: Clear liquid
Identity (IR-spectrum): Conforms
Water (H2O): 0.2% max

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Solvent effects on aromatic nucleophilic substitutions. Part 3. The kinetics of the reaction of 2, 4-dinitrochlorobenzene with piperidine in aprotic solvents
Mancini PME, et al.
J. Chem. Soc. Perkin Trans. II, (7), 1133-1138 (1984)
Thermal investigations of CoAPO materials prepared by using piperidine as a structure-directing agent
Rajic N, et al.
Thermochimica Acta, 351(1-2), 119-124 (2000)
Synthesis of FER zeolite with piperidine as structure-directing agent and its catalytic application
Chu Weifeng, et al.
Chinese Journal of Catalysis, 38(11), 1880-1887 (2017)
Omar F Luna et al.
Molecules (Basel, Switzerland), 21(11) (2016-11-18)
The deprotection step is crucial in order to secure a good quality product in Fmoc solid phase peptide synthesis. 9-Fluorenylmethoxycarbonyl (Fmoc) removal is achieved by a two-step mechanism reaction favored by the use of cyclic secondary amines; however, the efficiency

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service