Skip to Content
Merck
All Photos(4)

Documents

PP100

Sigma-Aldrich

IgG1 Antibody, Mouse

1.95 mg/mL, Chemicon®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

clone

monoclonal

Assay

≥95% (SDS-PAGE)

manufacturer/tradename

Chemicon®

concentration

1.95 mg/mL

technique(s)

ELISA: suitable

shipped in

wet ice

target post-translational modification

unmodified

General description

Purity: 95% or greater by SDS-PAGE.

Application

Mouse IgG1 Monoclonal Antibody control validated for use in ELISA.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Physical form

Format: Purified
Protein A purified
Purified immunoglobulin. Liquid in 100 mM Phosphate buffer, pH 7.0. Contains no preservative.

Storage and Stability

Maintain for 1 year at -20°C from date of shipment. Aliquot to avoid repeated freezing and thawing. For maximum recovery of product, centrifuge the original vial after thawing and prior to removing the cap.

Legal Information

CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Helen L Quasnichka et al.
Journal of immunological methods, 297(1-2), 133-141 (2005-03-22)
There is a need for a reliable assay for the quantification of collagen type I synthesis in the guinea pig, an important model for many connective tissue diseases. Procollagen type I C-terminal propeptide (PICP) is the established marker of type
TNF-alpha induces TGF-beta1 expression in lung fibroblasts at the transcriptional level via AP-1 activation.
Deborah E Sullivan,MaryBeth Ferris,Hong Nguyen,Elizabeth Abboud,Arnold R Brody
Journal of Cellular and Molecular Medicine null
S Chabot et al.
Journal of immunology (Baltimore, Md. : 1950), 162(11), 6819-6828 (1999-06-03)
IL-10, a cytokine with important anti-inflammatory properties, is generated within the CNS during neuroinflammation. The mechanism for its production is poorly understood. Since infiltrating lymphocytes come into close proximity with the macrophage-like cells of the CNS, the microglia, we have
Regulation of human beta-cell adhesion, motility, and insulin secretion by collagen IV and its receptor alpha1beta1.
Kaido, T; Yebra, M; Cirulli, V; Montgomery, AM
The Journal of Biological Chemistry null
Integrin-driven actin polymerization consolidates long-term potentiation.
Kramar, EA; Lin, B; Rex, CS; Gall, CM; Lynch, G
Proceedings of the National Academy of Sciences of the USA null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service