Skip to Content
Merck
All Photos(1)

Key Documents

PLCC06210

Millipore

Regenerated Cellulose Membrane Filter, 5 kDa NMWCO

Ultracel®, filter diam. 63.5 mm

Synonym(s):

Ultrafiltration Discs, 5 kDa NMW, Ultrafiltration membrane discs

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104903
eCl@ss:
32031602
NACRES:
NB.24

material

regenerated cellulose membrane

Quality Level

sterility

non-sterile

feature

hydrophilic

manufacturer/tradename

Ultracel®

technique(s)

protein clean-up: suitable (desalting)
protein purification: suitable (concentration)

diam.

63.5 mm

filter diam.

63.5 mm

filtration area

28.7 cm2

matrix

Ultracel®

pore size

5 kDa NMWCO

shipped in

ambient

Looking for similar products? Visit Product Comparison Guide

General description

Ultrafiltration membranes are either made of cellulose or polyethersulfone (PES). These membranes are used in diafiltration, either in centrifugal or pressure-driven devices such as the Amicon® Stirred Cell for efficient buffer exchange. The ultrafiltration membranes can be used for both sample concentration and buffer exchange. This minimizes sample transfers and reduces sample loss.

Application

Ultrafiltration Discs, 5 kDa NMW has been used:
  • to concentrate Acaudina molpadioidea hydrolysates
  • for fractionation of tilapia by-product (TB) protein hydrolysate
  • to desalt the reaction solution for the preparation of plant phenol-derived polymeric dyes

Legal Information

Amicon is a registered trademark of Merck KGaA, Darmstadt, Germany
ULTRACEL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Assessment on multilayer ultrafiltration membrane for fractionation of tilapia by-product protein hydrolysate with angiotensin I-converting enzyme (ACE) inhibitory activity
Roslan J, et al.
Separation and Purification Technology, 173, 250-257 (2017)
Modification of ACE-inhibitory peptides from Acaudina molpadioidea using the plastein reaction and examination of its mechanism
Jiang S, et al.
Food Bioscience, 26, 1-7 (2018)
Kyung Min Im et al.
Journal of visualized experiments : JoVE, (118) (2016-12-09)
Effective hair dyeing through in situ incubation of keratin hair with the products of fungal laccase-catalyzed polymerization of plant phenols has been previously demonstrated. However, the dyeing process takes a long time to complete compared to commercial hair-dyeing products. To

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service