Skip to Content
Merck
All Photos(1)

Key Documents

PC711

Sigma-Aldrich

Anti-CHIP Rabbit pAb

liquid, Calbiochem®

Synonym(s):

Anti-Carboxyl Terminus of Hcs70-Interacting Protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43

biological source

rabbit

Quality Level

antibody form

serum

antibody product type

primary antibodies

clone

polyclonal

form

liquid

contains

≤0.1% Sodium azide as preservative

species reactivity

Drosophila, hamster, rat, human, monkey, mouse

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
avoid repeated freeze/thaw cycles

isotype

IgG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Rabbit polyclonal antibody supplied as undiluted serum. Recognizes the ~35 kDa CHIP protein.
Recognizes the ~35 kDa CHIP protein. May also detect bands at ~43, ~51, and ~70 kDa corresponding to the mono-ubiquitinated, di-ubiquitinated, and dimeric forms of CHIP, respectively.
This Anti-CHIP Rabbit pAb is validated for use in Immunoblotting for the detection of CHIP.

Immunogen

a recombinant protein consisting of full-length human CHIP fused to a His•Tag sequence

Application

Immunoblotting (1:200-1:1000)

Immunofluorescence (1:250)

Immunoprecipitation (1:20)

Warning

Toxicity: Standard Handling (A)

Physical form

Undiluted serum.

Reconstitution

Following initial thaw, aliquot and freeze (-20°C).

Analysis Note

Positive Control
COS-7 cells

Other Notes

May also detect bands at 43, 51, and 70 kDa, wich correspond to the mono-ubiquitinated, di-ubiquitinated, and dimeric forms of CHIP, respectively. Antibody should be titrated for optimal results in individual systems.
Xu, W., et al. 2002. Proc. Natl. Acad. Sci. USA99, 12847.
Demand, J., et al. 2001. Curr. Biol.11, 1569.
Connell, P., et al. 2001. Nat. Cell Biol.3, 93.
Jiang. J., et al. 2001. J. Biol. Chem. 276, 42938.
Meacham, G.C., et al. 2001. Nat. Cell. Biol.3, 100.
Ballinger, C.A., et al. 1999. Mol. Cell. Biol.19, 4535.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Britney N Lizama et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 38(31), 6825-6840 (2018-06-24)
The C terminus of HSC70-interacting protein (CHIP, STUB1) is a ubiquitously expressed cytosolic E3-ubiquitin ligase. CHIP-deficient mice exhibit cardiovascular stress and motor dysfunction before premature death. This phenotype is more consistent with animal models in which master regulators of autophagy
Rudolf A Kley et al.
Brain : a journal of neurology, 135(Pt 9), 2642-2660 (2012-09-11)
Mutations in FLNC cause two distinct types of myopathy. Disease associated with mutations in filamin C rod domain leading to expression of a toxic protein presents with progressive proximal muscle weakness and shows focal destructive lesions of polymorphous aggregates containing
Jeannette N Stankowski et al.
Antioxidants & redox signaling, 14(10), 1787-1801 (2010-08-04)
The decision to remove or refold oxidized, denatured, or misfolded proteins by heat shock protein 70 and its binding partners is critical to determine cell fate under pathophysiological conditions. Overexpression of the ubiquitin ligase C-terminus of HSC70 interacting protein (CHIP)
Xinjia Wang et al.
Molecular endocrinology (Baltimore, Md.), 19(6), 1474-1482 (2005-03-12)
The ubiquitin/proteasome-dependent protein degradation pathway (UPP) is responsible for the accelerated down-regulation of glucocorticoid receptor (GR) levels in cells subjected to chronic glucocorticoid exposure. Whereas hormone-dependent down-regulation of GR operates in most cells, the receptor is not down-regulated after long-term
Nadja Kettern et al.
PloS one, 6(1), e16398-e16398 (2011-02-02)
The maturation of mouse macrophages and dendritic cells involves the transient deposition of ubiquitylated proteins in the form of dendritic cell aggresome-like induced structures (DALIS). Transient DALIS formation was used here as a paradigm to study how mammalian cells influence

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service