Skip to Content
Merck
All Photos(1)

Key Documents

OP10

Sigma-Aldrich

Anti-c-Myc (Ab-1) Mouse mAb (9E10)

liquid, clone 9E10, Calbiochem®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

9E10, monoclonal

form

liquid

contains

≤0.1% sodium azide as preservative

species reactivity

rat (weakly), human, mouse (weakly)

manufacturer/tradename

Calbiochem®

storage condition

do not freeze

isotype

IgG1

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... MYC(4609)
mouse ... Myc(17869)
rat ... Myc(24577)

General description

Purified mouse monoclonal antibody generated by immunizing BALB/c mice with the specified immunogen and fusing splenocytes with SP2/0 mouse myeloma cells. Recognizes the ~64-67 kDa (apparent MW) c-Myc protein.
Recognizes the ~60-67 kDa c-Myc protein in HL60 cells and lung carcinoma tissue.
This Anti-c-Myc (Ab-1) Mouse mAb (9E10) is validated for use in FC, Frozen Sections, Immunoblotting, IF, IP, Chromatin IP, Paraffin Sections for the detection of c-Myc (Ab-1).

Immunogen

Epitope: within amino acids 410-419
Human
a synthetic peptide (AEEQKLISEEDLLRKRREQLKHKLEQLRNSCA) corresponding to amino acids 408-439 of human c-Myc

Application

Flow Cytometry (20 µg/ml or use Cat. No. OP10F)

Frozen Sections (2-10 µg/ml)

Immunoblotting (1-5 µg/ml, see application references)

Immunofluorescence (1-5 µg/ml or use Cat. No. OP10F)

Immunoprecipitation (1 µg/sample)

Chromatin Immunoprecipitation (5 µg/ml, see application references)

Paraffin Sections (not recommended)

Packaging

Please refer to vial label for lot-specific concentration.

Warning

Toxicity: Standard Handling (A)

Physical form

In 0.05 M sodium phosphate buffer, 0.2% gelatin.

Analysis Note

Negative Control
HT1080 cells
Positive Control
HL-60 cells or lung carcinoma

Other Notes

LeGouy, E., et al. 1987. In Nuclear Oncogenes, Cold Spring Harbor Laboratory, 144.
Cole, M.D. 1986. Ann. Rev. Gen.20, 361.
Nisen, P.D., et al. 1986. Cancer Res.46, 6217.
Nau, M.M., et al. 1985. Nature318, 69.
Persson, H., et al. 1984. Science225, 687.
Alitalo, K., et al. 1983. Proc. Natl. Acad. Sci. USA80, 1707.
The c-Myc protein is extremely labile. c-Myc degradation can be as a result of freeze/thaw cycles, thus, we recommend using fresh lysates with a cocktail of protease inhibitors. c-Myc is often destroyed during the processing of paraffin sections and can therefore be difficult to detect in this application. This antibody recognizes the c-Myc protein and its cleavage products. The sequence of c-Myc predicts a 45 kDa protein, but c-Myc migrates under reducing conditions as a 64-67 kDa band. This antibody will not detect v-Myc but may react weakly to rodent c-Myc when used at high concentrations (10 µg/ml). An extra 34-40 kDa band may be detected in a immunoblot. May also be used in immunofluorescence as well as the detection and purification of recombinant proteins tagged with the Myc epitope sequence EQKLISEEDL (see application references). The immunogen, c-Myc (Peptide-1) is also available for competition studies (Cat. No. PP06). Antibody should be titrated for optimal results in individual systems.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

L A Cass et al.
Molecular and cellular biology, 19(9), 5882-5891 (1999-08-24)
The effects of cyclic AMP (cAMP) on cell proliferation are cell type specific. Although the growth-inhibitory effects of cAMP have been well studied, much less is known regarding how cAMP stimulates proliferation. We report that cAMP stimulates proliferation through both
Ling Zhu et al.
The Plant cell, 28(4), 855-874 (2016-04-14)
The phytochrome interacting factors (PIFs), a small group of basic helix-loop-helix transcription factors, repress photomorphogenesis both in the dark and light. Light signals perceived by the phytochrome family of photoreceptors induce rapid degradation of PIFs to promote photomorphogenesis. Here, we
Luciano Mazzoccoli et al.
Oncology reports, 42(2), 775-784 (2019-06-08)
Deregulation of microRNA expression plays a significant role in several cancer types including Burkitt lymphoma (BL). MicroRNA genes may be regulated through epigenetic mechanisms, such as specific histone modifications and/or DNA methylation of CpG islands in promoter regions, or by
David R Simpson et al.
Cancer research, 71(11), 3822-3830 (2011-05-26)
c-Myc is an oncogene transcription factor that causes cancer in many settings, including solid tumors that arise in the context of organized tissue structures. Given that disruption of tissue architecture frequently occurs in cancer, there is considerable interest in how
Meleah A Hickman et al.
PLoS genetics, 5(11), e1000710-e1000710 (2009-11-07)
Deacetylases of the Sir2 family regulate lifespan and response to stress. We have examined the evolutionary history of Sir2 and Hst1, which arose by gene duplication in budding yeast and which participate in distinct mechanisms of gene repression. In Saccharomyces

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service