Skip to Content
Merck

MATHKSA

NEP Control HKSA- PyroMAT® System MAT

Non-endotoxin pyrogen control of heat killed Staphyloccus aureus for use in monocyte activation test

Synonym(s):

NEP, NEP control

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116130
Suitability:
pyrogenic bacteria
Manufacturer/tradename:
PyroMAT®
Application(s):
pharmaceutical
pyrogen testing
Storage temp.:
2-8°C
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

product line

PyroMAT®

feature

Non-endotoxin pyrogen control of heat killed Staphyloccus aureus for use in monocyte activation test

packaging

pkg of 1 vial

manufacturer/tradename

PyroMAT®

application(s)

pharmaceutical
pyrogen testing

compatibility

configured for PyroDetect System, configured for PyroMAT® System

shipped in

ambient

storage temp.

2-8°C

suitability

pyrogenic bacteria

General description

The PyroMAT® System uses cryopreserved Mono-Mac-6 (MM6) human monocytic cells as a source of monocytes. The response to a full range of pyrogens, including endotoxins and non-endotoxin pyrogens (NEPs)is determined by the measurement of interleukin-6 (IL-6) produced by the Mono-Mac-6 cells using ELISA.
NEP control HKSA is a heat-killed preparation of Staphylococcus aureus, supplied in a lyophilized form. HKSA is a non-endotoxin pyrogen (NEP) that activates the TLR2 receptor of monocytes and can be used as a positive control with the PyroMAT® System (Monocyte Activation Test for pyrogen detection).

Application

NEP control of heat-killed Staphylococcus aureus can be used as a positive control with the PyroMAT® System. It is to be used for Product Specific Validation (PSV) and routine testing to show that the system can detect non-endotoxin pyrogen contaminants in pharmaceutical products.

Biochem/physiol Actions

TLR-2 agonist

Features and Benefits

Endotoxin-free product at concentration of use in MAT

Preparation Note

Store unopened vials at 2-8°C
Do not use unopened vials after expiration date
Store resuspended aliquots at -20°C
Shelf life of aliquots can be found on the technical datasheet

Legal Information

PYROMAT is a registered trademark of Merck KGaA, Darmstadt, Germany


pictograms

Exclamation mark

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

flash_point_f

Not applicable

flash_point_c

Not applicable

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral



Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library