Skip to Content
Merck
All Photos(1)

Key Documents

MABX49

Sigma-Aldrich

Anti-Chikungunya (CHIKV-Env 1 & 2)

Synonym(s):

Anti-CHIKV Antibody

Sign Into View Organizational & Contract Pricing

Select a Size

10 EA
€146.00
5 EA
€158.00
1 EA
€208.00

€146.00


Please contact Customer Service for Availability

To order products, please contact your local dealer.

Select a Size

Change View
10 EA
€146.00
5 EA
€158.00
1 EA
€208.00

About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

€146.00


Please contact Customer Service for Availability

To order products, please contact your local dealer.

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX49-1KC)
pkg of 10 mg (MABX49-10KC)

technique(s)

ELISA: suitable
immunodiffusion: suitable

isotype

IgG1

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
GF98104282GF19711908GF34444997
assay

99%

assay

99%

assay

99%

assay

99%

manufacturer/tradename

Goodfellow 914-681-87

manufacturer/tradename

Goodfellow 981-042-82

manufacturer/tradename

Goodfellow 197-119-08

manufacturer/tradename

Goodfellow 344-449-97

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

density

2.7 g/mL at 25 °C (lit.)

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

resistivity

2.6548 μΩ-cm

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

bp

2460 °C (lit.)

Specificity

Chikungunya Virus Env 1 & 2. No reactivity with Dengue, Zika, WNV, TBV or JEV.

Immunogen

Recombinant Chikungunya Virus Envelope Protein

Application

This mouse monoclonal Anti-Chikungunya (CHIKV-Env 1 & 2), Cat. No. MABX49, is tested for use in ELISA and Lateral Flow Assay, for the detection of Recombinant Chikungunya Virus Envelope Protein (CHIKV E1 and E2).

Target description

Chikungunya virus is an arthropod-borne virus (arbovirus) spread by the bite of infected Aedes mosquitoes.  It is a member of the Alphavirus genus of RNA viruses.  Symptoms of fever and joint pain usually appear within a week of infection.  Muscle pain, headache, fatigue and rash may also occur.

Linkage

Recommended Matched Pairs MABX49 with MABX50 and/or MABX51.

Physical form

Purified mouse monoclonal in buffer containing 0.015 M Potassium Phosphate, pH 7.2 with 0.85% Sodium Chloride and 0.05% Sodium Azide

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


  • Certificates of Analysis (COA)

    Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

    Already Own This Product?

    Find documentation for the products that you have recently purchased in the Document Library.

    Visit the Document Library

    Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

    Contact Technical Service