Skip to Content
Merck
All Photos(1)

Key Documents

MABX4182

Sigma-Aldrich

Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 182

clone 182, from mouse

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

182, monoclonal

species reactivity

human

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG2b

UniProt accession no.

Gene Information

human ... IGFBP4(3487)

General description

Antibody recognizes the C-term of Human insulin like growth factor binding protein 4 (IGFBP-4). IGFBP-4 is a member of the Insulin like growth factor family and is responsible for binding insulin like growth factors I and II. Research has linked levels of C-term (CT-IGFBP-4) and N-term (NT-IGFBP-4) to certain cardiac events, offering potential as a novel cardiac biomarker

Application

Confirmed matched pairing. Capture Antibody MABX4182-1KC; Detector Antibody MABX4144-1KC or Detector Antibody MABX4163-1KC
Detect CT-IGFBP-4 using this mouse monoclonal Anti- Insulin-like growth factor binding protein 4 (IGFBP-4) clone 182, Cat. No. MABX4182-1KC, validated for use in Western Blotting and ELISA.

Quality

Tested

Physical form

Format: Purified

Other Notes

Concentration: Please refer to lot specific datasheet.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service