Skip to Content
Merck
All Photos(1)

Key Documents

MABX41

Sigma-Aldrich

Anti-Mycoplasma pneumoniae

Synonym(s):

Anti-M. pneumonia Antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified antibody

clone

monoclonal

form

solution

species reactivity

all

packaging

pkg of 1 mg (MABX41-1KC)
pkg of 10 mg (MABX41-10KC)

technique(s)

immunodiffusion: suitable

isotype

IgG2a

relevant disease(s)

infectious diseases

shipped in

wet ice

storage temp.

2-8°C

Specificity

M.pneumoniae P30. No reactivity with M. fermentans, M. genitalium, M. salvarium, M. hominis, or M. orlae.

Immunogen

M. pneumoniae FH

Application

This mouse monoclonal antibody, Cat. No. MABX41, is tested for use in Lateral Flow Assay for the detection of M. pneumonia P30.

Target description

Mycoplasma pneumoniae is a type of "atypical" bacteria that commonly causes up to one-fifth of all lung infections acquired outside the hospital.  M. pneumoniae is very small and characterized by the absence of a peptidoglycan cell wall and resulting in resistance to many antibacterial agents. The persistence of M. pneumoniae infection even after treatment is associated to the ability to mimic host cell surface composition.

Linkage

Recommended Matched Pair MABX41 capture and MABX40 detection

Physical form

Purified mouse monoclonal in buffer containing 10 mM Phosphate Buffered Saline, pH 7.4 with 0.05% Sodium Azide

Reconstitution

Please refer to Certificate of Analysis for concentration.

Storage and Stability

Store at 2-8 °C. For long-term storage, freeze at -20 °C

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

related product

Product No.
Description
Pricing

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service