Skip to Content
Merck
All Photos(1)

Documents

MABS274

Sigma-Aldrich

Anti-LRP6, clone A59, Ectodomain Antibody

clone A59, from mouse

Synonym(s):

Low-density lipoprotein receptor-related protein 6, LRP-6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

A59, monoclonal

species reactivity

human

technique(s)

flow cytometry: suitable
immunocytochemistry: suitable
immunoprecipitation (IP): suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... LRP6(4040)

General description

Low density lipoprotein receptor-related protein 6, also known as LRP6, is a member of the low density Lipoprotein receptor gene family (LDLR). LDLR are cell surface proteins. LRP6 plays a crucial role in embryonic development by functioning as a co-receptor for the Wnt signaling pathway. LRP6 is highly homologous and presents itself as a dimer in the cell membrane until it is phosphlorylated by Wnt. LRP6 has a high affinity with the Dickkopf (DKK) family of antagonists which, when bound, block Wnt. When LRP6 binds to Wnt it is phosphlorated at multiple sites, which results in Axin, a scaffolding protein, being recruited to the cell membrane and β-catenin activation. β-catenin prevents Wnt and LRP6 from binding. LRP6 is associated with autosomal dominant coronary artery disease type 2. Mutations resulting in the omission of the extracellular domain of LPR6 results in constant activity whereas carboxy intracellular domain exclusion has a negative effect, resulting in the blocking of Wnt signaling.

Immunogen

Epitope: Ectodomain Domain
Purified protein corresponding to the ectodomain domain of human LRP6.

Application

Flow Cytometry Analysis: A representative lot from an independent laboratory detected LRP6, Ectodomain in Chimeric LRP6 expressing 293T cells (Yasui, N., et al. (2010). J Immunol Methods. 352(1-2):153-160.).

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated LRP6, Ectodomain from Wnt3a-treated HEK293T cell lysate (Yasui, N., et al. (2010). J Immunol Methods. 352(1-2):153-160.).

Immunocytochemistry Analysis: A representative lot from an independent laboratory detected LRP6, Ectodomain in HeLa S3 cells (Yasui, N., et al. (2010). J Immunol Methods. 352(1-2):153-160.).
Research Category
Signaling
Research Sub Category
Insulin/Energy Signaling
This Anti-LRP6 antibody, clone A59, Ectodomain is validated for use in flow cytometry, IP & ICC for the detection of LRP6.

Quality

Evaluated by Flow Cytometry in HEK293 cells.

Flow Cytometry Analysis: 2 µg of this antibody detected LRP6, Ectodomain in 1X10E6 HEK293 cells.

Target description

180 kDa calculated

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bhaven B Patel et al.
PloS one, 14(1), e0198463-e0198463 (2019-01-30)
The systematic identification of regulatory elements that control gene expression remains a challenge. Genetic screens that use untargeted mutagenesis have the potential to identify protein-coding genes, non-coding RNAs and regulatory elements, but their analysis has mainly focused on identifying the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service