Skip to Content
Merck
All Photos(1)

Key Documents

MABN42

Sigma-Aldrich

Anti-Neuregulin-1 Antibody, clone D11

clone D11, from mouse

Synonym(s):

neuregulin 1, sensory and motor neuron derived factor, neu differentiation factor, pro-neuregulin-1, membrane-bound isoform, heregulin, alpha (45kD, ERBB2 p185-activator), heregulin, alpha (45kD, ERBB2 p185-activator), glial growth factor

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
D11, monoclonal
application:
ICC
WB
species reactivity:
mouse
technique(s):
immunocytochemistry: suitable
western blot: suitable
citations:
2

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

D11, monoclonal

species reactivity

mouse

technique(s)

immunocytochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

mouse ... Nrg1(211323)

General description

The group of Neuregulin-1 proteins are cell-cell signaling molecules and are ligands for receptor tyrosine kinases of the ErbB/HER subfamily. Type III NRG1 proteins (also referred to as CRD-NRG1 proteins) play a critical role in neural development, including myelination, motor and sensory neuron survival, and neuromuscular synapse development (Falls, 2003). Defects in animals lacking type III NRGs include retraction of nerve terminals from newly formed synapses, absence of Schwann cells from peripheral nerves, and loss of motor and sensory neurons (Wolpowitz et al., 2000). Available evidence indicates that most Type III NRG1 proteins are synthesized as transmembrane proteins with 2 TM domains, and that proteolytic processing produces a bioactive transmembrane form (NTFm) which serves as a juxtacrine signal (Wang, 2001; Falls, 2003).

Specificity

This antibody recognizes Neuregulin-1 at the extracellular domain.

Immunogen

Epitope: Extracellular domain
His-tagged recombinant protein corresponding to mouse Neuregulin-1 at the extracellular domain.

Application

Detect Neuregulin-1 using this Anti-Neuregulin-1 Antibody, clone D11 validated for use in WB & IC.
Immunocytochemistry Analysis: A previous lot of this antibody was used successfully by an independent laboratory in IC. (Pirotte, D., et al.)
Research Category
Neuroscience
Research Sub Category
Developmental Neuroscience

Quality

Evaluated by Western Blot in mouse brain microsomal tissue lysate.

Western Blot Analysis: 0.25 µg/mL of this antibody detected Neuregulin-1 on 10 µg of mouse brain microsomal tissue lysate.

Target description

~29 kDa observed. This target has been shown to undergo differential splicing during development into ~30 different isoforms.

Physical form

Format: Purified
Protein G
Purified mouse monoclonal IgG1κ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
Mouse brain microsomal tissue lysate

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ki H Ma et al.
Glia, 66(11), 2487-2502 (2018-10-12)
The transition of differentiated Schwann cells to support of nerve repair after injury is accompanied by remodeling of the Schwann cell epigenome. The EED-containing polycomb repressive complex 2 (PRC2) catalyzes histone H3K27 methylation and represses key nerve repair genes such
Erin-Mai F Lim et al.
Proceedings of the National Academy of Sciences of the United States of America, 114(9), E1707-E1716 (2017-02-01)
AlphaB-crystallin (αBC) is a small heat shock protein that is constitutively expressed by peripheral nervous system (PNS) axons and Schwann cells. To determine what role this crystallin plays after peripheral nerve damage, we found that loss of αBC impaired remyelination

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service