MABN293
Anti-Peripherin-2 Antibody, clone 6B10.1
clone 6B10.1, from mouse
Synonym(s):
Peripherin-2, Retinal degeneration slow protein, Tetraspanin-22, Tspan-22
About This Item
biological source
mouse
Quality Level
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
6B10.1, monoclonal
species reactivity
mouse, rat, human
technique(s)
immunohistochemistry: suitable
western blot: suitable
isotype
IgG1κ
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PRPH2(5961)
General description
Immunogen
Application
Neuroscience
Developmental Signaling
Immunohistochemistry Analysis: A 1:2,000 dilution from a representative lot detected Peripherin-2 in human retina tissue.
Quality
Western Blotting Analysis: A 1:1,000 dilution of this antibody detected Peripherin-2 in 10 µg of mouse eye tissue lysate.
Target description
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service