Skip to Content
Merck
All Photos(2)

Key Documents

MABN1190

Sigma-Aldrich

Anti-DLG4 Antibody

mouse monoclonal, 1A8.1

Synonym(s):

Disks large homolog 4, Postsynaptic density protein 95, PSD-95, Synapse-associated protein 90, SAP-90, SAP90

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

Product Name

Anti-PSD95 Antibody, clone 1A8.1, clone 1A8.1, from mouse

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

1A8.1, monoclonal

species reactivity

rat, human

technique(s)

immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

NCBI accession no.

UniProt accession no.

target post-translational modification

unmodified

Gene Information

human ... DLG4(1742)

General description

Disks large homolog 4 (UniProt P78352; also known as Postsynaptic density protein 95, PSD-95, SAP-90, SAP90, Synapse-associated protein 90) is encoded by the DLG4 (also known as PSD95) gene (Gene ID 1742) in human. PSD-95 belong to the family of membrane-associated guanylate kinases (MAGUKs), which also include PSD-93, SAP97 and SAP102. PSD95 is predominantly expressed in the hippocampus CA1 region and prefrontal cortex of the brain, where it localizes in a somatodendritic pattern in the post-synaptic membrane and in presynaptic axon terminals of inhibitory neurons. PDS-95 is the major scaffolding protein in the excitatory postsynaptic density (PSD) and a potent regulator of synaptic strength via interactions with a diverse PSD proteins, including NMDA receptors (NMDARs), AMPA receptor (AMPAR) complexes via Stargazin/TARP, adhesion molecules, as well as other scaffolding proteins, such as GKAP and Shank. PSD95 and neuronal nitric-oxide synthase (nNOS) interaction plays a role in maintaining hypersensitivity in acute and chronic pain. In addition, the assembly of the GluR6-PSD95-CaMKII complex is implicated as a majore signaling module that mediates injury induced by brain ischemia by facilitating GluR6 serine phosphorylation by CaMKII and the activation of JNK.

Specificity

Clone 1A8.1 epitope sequence is present in all spliced isoforms of human, mouse, and rat PSD95 reported by UniProt (P78352, Q62108, P31016).

Immunogen

Epitope: Region between PDZ domain 2 & 3.
KLH-conjugated linear peptide corresponding to the region between PDZ domain 2 & 3 of human PSD95.

Application

Anti-PSD95 Antibody, clone 1A8.1 is an antibody against PSD95 for use in Western Blotting, Immunohistochemistry.
Immunohistochemistry Analysis: A 1:50 dilution from a representative lot PSD95 in human cerebral cortex and cerebellum tissue sections.
Research Category
Neuroscience
Research Sub Category
Synapse & Synaptic Biology

Quality

Evaluated by Western Blotting in rat hippocampus tissue lysate.

Western Blotting Analysis: 0.5 µg/mL of this antibody detected PSD95 in 10 µg of rat hippocampus tissue lysate.

Target description

~80 kDa observed. 80.50/80.47/80.47 kDa human/mouse/rat isoform 1; PSD95-alpha), 85.43/85.42 kDa (human/mouse isoform 2; PSD95-beta), 80.13/80.10 kDa (human/mouse isoform 3) calculated.

Physical form

Format: Purified
Protein G purified.
Purified mouse monoclonal IgG1κ antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Beatrice Vignoli et al.
Cells, 11(9) (2022-05-15)
Neurons release and respond to brain-derived neurotrophic factor (BDNF) with bursts of brain activity. BDNF action is known to extend to peri-synaptic astrocytes, contributing to synaptic strengthening. This implies that astrocytes have a set of dynamic responses, some of which
Angela O Dorigatti et al.
GeroScience, 43(1), 115-124 (2020-10-17)
The ability to generate in vitro cultures of neuronal cells has been instrumental in advancing our understanding of the nervous system. Rodent models have been the principal source of brain cells used in primary cultures for over a century, providing
Daozhong Jin et al.
Journal of neurochemistry, 164(2), 143-157 (2022-10-13)
Glutamate NMDA receptors (NMDARs) in the nucleus accumbens (NAc) are critically involved in drug dependence and reward. α2δ-1 is a newly discovered NMDAR-interacting protein that promotes synaptic trafficking of NMDARs independently of its conventional role as a calcium channel subunit.
Yuying Huang 黄玉莹 et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 44(4) (2023-12-21)
K+-Cl- cotransporter-2 (KCC2) critically controls neuronal chloride homeostasis and maintains normal synaptic inhibition by GABA and glycine. Nerve injury diminishes synaptic inhibition in the spinal cord via KCC2 impairment. However, how KCC2 regulates nociceptive input to spinal excitatory and inhibitory
Daozhong Jin et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 43(31), 5593-5607 (2023-07-15)
Aberrant activation of presynaptic NMDARs in the spinal dorsal horn is integral to opioid-induced hyperalgesia and analgesic tolerance. However, the signaling mechanisms responsible for opioid-induced NMDAR hyperactivity remain poorly identified. Here, we show that repeated treatment with morphine or fentanyl

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service