biological source
mouse
Quality Level
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
F2.67, monoclonal
mol wt
calculated mol wt 13.53 kDa
purified by
using protein G
species reactivity
mouse
packaging
antibody small pack of 100 μL
technique(s)
flow cytometry: suitable
isotype
IgG2aκ
epitope sequence
Unknown
UniProt accession no.
shipped in
2-8°C
target post-translational modification
unmodified
Related Categories
General description
Specificity
Immunogen
Application
Evaluated by Flow Cytometry in Balb/c mouse thymocytes.
Flow Cytometry Analysis: 1.0 μg of this antibody detected T-cell Vg 7 in one million Balb/c mouse thymocytes.
Tested Applications
Flow Cytometry Analysis: A representative lot detected TCR Vg7 in Flow Cytometry applications (Pereira, P., et al. (2000). Eur J Immunol.;30(7):1988-97; Dalton, J.E., et al. (2006). Gastroenterology.;131(3):818-29; Newton, D.J., et al. (2006). Infect Immun.;74(2):1097-105; Hamada, S. et al. (2008). Immunology.;125(2):170-7; Li, C., et al. (2012). Cell Immunol.;279(2):151-9; Walker, C.R., et al. (2013). PLoS One.;8(12):e84553).
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Physical form
Storage and Stability
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service