Skip to Content
Merck
All Photos(1)

Key Documents

MABF144B

Sigma-Aldrich

Anti-KLRG1 (mouse), violetFluor® 450, clone 2F1 Antibody

clone 2F1, from hamster(Syrian), violetFluor® 450

Synonym(s):

Killer cell lectin-like receptor subfamily G member 1, Mast cell function-associated antigen 2F1, KLRG1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

hamster (Syrian)

Quality Level

conjugate

violetFluor® 450

antibody form

purified antibody

antibody product type

primary antibodies

clone

2F1, monoclonal

species reactivity

mouse

packaging

antibody small pack of 25 μg

technique(s)

flow cytometry: suitable

isotype

IgG

UniProt accession no.

target post-translational modification

unmodified

Gene Information

mouse ... Klrg1(50928)

General description

The 2F1 antibody reacts with mouse KLRG1 (Killer cell Lectin-like Receptor G1). This 30-38 kDa homodimeric receptor may be expressed by activated, mature NK cells and by effector/memory T cells, with potentially different roles in each cell type. KLRG1 can regulate, in an inhibitory fashion, the development and effector functions of NK cells, and is often cited as a senescence or terminal differentiation marker for T cells. Ligands for KLRG1 include members of the cadherin family of adhesion molecules, specifically N-Cadherin, E-Cadherin, and R-Cadherin. These interactions may induce bidirectional, immunosuppressive signaling in both KLRG- and Cadherin-expressing cells. A more recently identified role for KLRG1-Cadherin signaling in tissue organization, e.g. in cardiac angiogenesis, expands the function of these interactions beyond immunosuppression of immune cells. (Bouchentouf et al. 2010. J. Immunol. 185: 7014-7025).

Immunogen

Immunogen information for this antibody is proprietary information.

Application

Anti-KLRG1 (mouse), violetFluor® 450, clone 2F1 Antibody is an antibody against KLRG1 for use in Flow Cytometry.
Research Category
Inflammation & Immunology
Research Sub Category
Immunoglobulins & Immunology

Quality

This monoclonal antibody was purified from tissue culture supernatant via affinity chromatography. The purified antibody was conjugated under optimal conditions, with unreacted dye removed from the preparation.

Flow Cytometry Analysis: 0.125 μg from a representative lot detected KLRG1 in one million C57Bl/6 splenocytes stained with PE Anti-mouse NK1.1.

Physical form

Purified Golden Syrian Hamster monoclonal containing 10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2.
Purified by size-exclusion chromatography

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: It is recommended to store the product undiluted at 2-8°C and protected from prolonged exposure to light. Do not freeze.

Other Notes

Concentration: Please refer to lot specific datasheet.

Legal Information

violetFluor is a registered trademark of Tonbo Biotechnologies

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service