Skip to Content
Merck
All Photos(2)

Key Documents

MABE911

Sigma-Aldrich

Anti-Aiolos Antibody, clone 9D10

clone 9D10, from mouse

Synonym(s):

Zinc finger protein Aiolos, Ikaros family zinc finger protein 3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9D10, monoclonal

species reactivity

human, mouse

technique(s)

ChIP: suitable (ChIP-seq)
western blot: suitable

isotype

IgG2bκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... IKZF3(22806)

General description

Aiolos is a zinc finger DNA binding transcription factor important in lymphocyte differentiation and development. Aiolos plays an essential role in the regulation of B-cell differentiation including, proliferation and maturation to plasma cells. Aiolos also works as a cofactor in gene suppression via its interaction with the transcription factor FOXP3 in regulatory T cells as well, and Aiolos is involved in various chromatin functions too through its interaction with histone deacetylase complexes containing HDAC1, MTA2 and SIN3A. Aiolos is localized in the nucleus primarily but can be found in the cytoplasm depending upon the cellular situation. Aiolos is expressed in leukocytes and found in high concentration in spleen and thymus and has multiple isoforms. Activity is regulated via phosphorylation.

Immunogen

Recombinant protein corresponding to human Aiolos.

Application

Chromatin Immunoprecipitation (ChIP): A representative lot of this antibody was reported to work in ChIP (see Zhang, J., et al 2012).

ChIP-seq: A representative lot of this antibody was reported to work in ChIP (see Zhang, J., et al 2012).

Western Blotting Analysis: A 1 µg/mL dilution of this antibody detected Aliolos in an HL-60 cell lysate
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Transcription Factors
This Anti-Aiolos Antibody, clone 9D10 is validated for use in Western Blotting and ChIP-seq and Chromatin Immunoprecipitation (ChIP) for the detection of Aiolos.

Quality

Evaluated by Western Blotting in a Jurkat cell lysate.

Western Blotting Analysis: A 1 µg/mL dilution of this antibody detected Aliolos in 10 µg of a Jurkat cell lysate.

Target description

~ 58-70 kDa observed

Physical form

Format: Purified
Protein G Purified
Purified mouse monoclonal IgG2bκ in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gregory Lazarian et al.
Cancer cell, 39(3), 380-393 (2021-03-11)
Hotspot mutation of IKZF3 (IKZF3-L162R) has been identified as a putative driver of chronic lymphocytic leukemia (CLL), but its function remains unknown. Here, we demonstrate its driving role in CLL through a B cell-restricted conditional knockin mouse model. Mutant Ikzf3
Jingjie Chang et al.
Frontiers in immunology, 14, 1239779-1239779 (2023-09-04)
AIOLOS, encoded by IKZF3, is a member of the IKZF family of proteins that plays an important role in regulating late B-cell differentiation. Human individuals heterozygous for the AIOLOS p.N160S variant displayed impaired humoral immune responses as well as impaired
Hye Sun Kuehn et al.
The Journal of experimental medicine, 218(12) (2021-10-26)
AIOLOS/IKZF3 is a member of the IKAROS family of transcription factors. IKAROS/IKZF1 mutations have been previously associated with different forms of primary immunodeficiency. Here we describe a novel combined immunodeficiency due to an IKZF3 mutation in a family presenting with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service