Skip to Content
Merck
All Photos(2)

Key Documents

MABE647

Sigma-Aldrich

Anti-acetyl-Histone H3 (Lys27) Antibody, clone RM172

clone RM172, from rabbit

Synonym(s):

Histone H3.3, Histone H3

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

biological source

rabbit

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

RM172, monoclonal

species reactivity

human

technique(s)

multiplexing: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

target post-translational modification

acetylation (Lys27)

Gene Information

human ... H3F3B(3021)

General description

Histone H3, also known as Histone H3.1t (H3/t), H3t, H3/g, and encoded by the gene name HIST3H3/ H3FT, is a core component of nucleosome. Nucleosomes wrap and compact DNA into chromatin, limiting DNA accessibility to the cellular machineries which require DNA as a template. Therefore, histones play a central role in transcription regulation, DNA repair, DNA replication and chromosomal stability. Featuring a main globular domain and a long N-terminal tail, H3 is involved with the structure of the nucleosomes of the ′beads on a string′ structure. The N-terminal tail of histone H3 protrudes from the globular nucleosome core and can undergo several different types of epigenetic modifications that influence cellular processes. These modifications include the covalent attachment of methyl or acetyl groups to lysine and arginine amino acids and the phosphorylation of serine or threonine. High levels of phosphorylation of Histone H3 are associated with mitosis.

Specificity

This antibody reacts to Histone H3 acetylated at Lysine 27 (K27ac). No cross reactivity with other acetylated Lysines in Histone H3.

Immunogen

A linear peptide corresponding to human Histone H3 acetylated at Lys27.

Application

Multiplex Analysis (Mplex): 0.1-0.5 μg/mL from a representative lot was used in Multiplex.
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Chromatin Biology
This Anti-acetyl-Histone H3 (Lys27) Antibody, clone RM172 is validated for use in Western Blotting, Multiplexing for the detection of acetyl-Histone H3.

Quality

Evaluated by Western Blotting in untreated and sodium butyrate treated HeLa acid extract.

Western Blotting Analysis (WB): 1 μg/mL of this antibody detected acetyl-Histone H3 (Lys27) in sodium butyrate treated HeLa acid extract.

Target description

~17 kDa observed. Uncharacterized bands may be observed in some lysate(s).

Physical form

Format: Purified
Protein A purified
Rabbit monoclonal in PBS with 1% BSA and 0.09% sodium azide

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.
Note: Variability in freezer temperatures below -20°C may cause glycerol containing solutions to become frozen during storage.

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Michael S Hoetker et al.
Nature cell biology, 25(8), 1121-1134 (2023-07-18)
The epigenetic mechanisms that maintain differentiated cell states remain incompletely understood. Here we employed histone mutants to uncover a crucial role for H3K36 methylation in the maintenance of cell identities across diverse developmental contexts. Focusing on the experimental induction of
Chi Sun et al.
Scientific reports, 13(1), 12899-12899 (2023-08-10)
Enhancers function with a basal promoter to control the transcription of target genes. Enhancer regulatory activity is often studied using reporter-based transgene assays. However, unmatched results have been reported when selected enhancers are silenced in situ. In this study, using
Steven J Wu et al.
Nature biotechnology, 39(7), 819-824 (2021-04-14)
Methods for quantifying gene expression1 and chromatin accessibility2 in single cells are well established, but single-cell analysis of chromatin regions with specific histone modifications has been technically challenging. In this study, we adapted the CUT&Tag method3 to scalable nanowell and
Yaw Asante et al.
Nature communications, 14(1), 8361-8361 (2023-12-16)
Activation of oncogenic gene expression from long-range enhancers is initiated by the assembly of DNA-binding transcription factors (TF), leading to recruitment of co-activators such as CBP/p300 to modify the local genomic context and facilitate RNA-Polymerase 2 (Pol2) binding. Yet, most
Michael P Meers et al.
eLife, 8 (2019-06-25)
Previously, we described a novel alternative to chromatin immunoprecipitation, CUT&RUN, in which unfixed permeabilized cells are incubated with antibody, followed by binding of a protein A-Micrococcal Nuclease (pA/MNase) fusion protein (Skene and Henikoff, 2017). Here we introduce three enhancements to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service