Skip to Content
Merck
All Photos(1)

Key Documents

MABE290

Sigma-Aldrich

Anti-Estrogen Receptor β Antibody, clone 9.88

clone 9.88, from mouse

Synonym(s):

Estrogen receptor beta, ER-beta, Nuclear receptor subfamily 3 group A member 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41
clone:
9.88, monoclonal
application:
IP
WB
species reactivity:
human, mouse, rat
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:
1

biological source

mouse

Quality Level

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

9.88, monoclonal

species reactivity

human, mouse, rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgMκ

NCBI accession no.

UniProt accession no.

shipped in

wet ice

target post-translational modification

unmodified

Gene Information

human ... ESR2(2100)

General description

Estrogen receptor beta (ERβ; NR3A2 or ESR2) is a nuclear steroid receptor that has been detected in the ovary, testis, thymus, spleen, bone, brain, and kidney. It is a multidomain protein containing A/B, C, D, E, and F domains, and it is activated by estrogen, resulting in homo- or heterodimerization (with ERα). The active ERβ-estrogen complex binds to regulatory regions known as estrogen response elements on target genes; however, ERβ may also regulate transcription without a bound ligand via its activation function (AF-1) region in the A/B domain. The ERβ receptor interacts with a wide variety of proteins including transcriptional corepressors and coactivators that regulate its activity. Its expression is also regulated by hypermethylation of CpG islands in the ERβ promoter. Previous studies have reported that ERβ has anti-inflammatory and anti-proliferative effects, and may therefore be an important therapeutic target for cancers, especially those occurring in the reproductive organs.

Immunogen

Recombinant protein corresponding to human Estrogen Receptor β.

Application

Immunoprecipitation Analysis: A representative lot from an independent laboratory immunoprecipitated Estrogen Receptor β in HUVEC cells (Simoncini, T., et al. (2006). Mol Endocrinol. 20(8):1756-1771.).
Research Category
Epigenetics & Nuclear Function
Research Sub Category
Cell Cycle, DNA Replication & Repair
Use Anti-Estrogen Receptor β Antibody, clone 9.88 (Mouse Monoclonal Antibody) validated in WB, IP to detect Estrogen Receptor β also known as Estrogen receptor beta.

Quality

Evaluated by Western Blot in PC-12 cell lysate.

Western Blot Analysis: A 1:1,000 dilution of this antibody detected Estrogen Receptor β in PC-12 cell lysate.

Target description

~55 kDa observed. Uniprot describes eight different isoforms ranging from ~36 kDa to ~59 kDa

Physical form

Format: Purified
Purified mouse monoclonal IgMκ in buffer containing PBS with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Analysis Note

Control
PC-12 cell lysate

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service